Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00295585_m1
          See other TCEB3-AS1 GE Assays ›
          Gene Symbol:
          TCEB3-AS1
          Gene Name:
          TCEB3 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 23760382 - 23778297 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00295585_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TCEB3-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038280.1 1 - 2 99 74
          GenBank mRNA AL049675.1 - 1 - 2 89 74
          BG429664.1 - 1 - 2 99 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013300.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002349.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 106877 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 357374 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 90951 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 117714 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 27832 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 260348 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 957178 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00030248 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P15057526 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00030250 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 85206 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15057518 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00030252 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00030244 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15057535 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00030248 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00030247 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00030250 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15057530 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15057526 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00030247 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00030248 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00030250 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P15057526 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01036188.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036188.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036188.hg.1 TC01002349.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01036191.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036191.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036191.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036191.hg.1 TC01002349.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01036192.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036192.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036192.hg.1 TC01002349.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01036193.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036193.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036193.hg.1 TC01002349.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036190.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036190.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01036190.hg.1 NR_038280.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01036190.hg.1 TC01002349.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100198612.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198612.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198612.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198612.hg.1 TC0100013300.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198612.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198612.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198612.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198606.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198606.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198606.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198606.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198606.hg.1 TC0100013300.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198606.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198606.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198606.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198606.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198614.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198614.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198614.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198614.hg.1 TC0100013300.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198614.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198614.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198614.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198609.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198609.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198609.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198609.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198609.hg.1 TC0100013300.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198609.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198609.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198609.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198609.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 ENST00000447961.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013300.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198602.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198602.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198602.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198602.hg.1 TC0100013300.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198602.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198602.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198602.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198610.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198610.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198610.hg.1 NR_038280.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100198610.hg.1 TC0100013300.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100198610.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198610.hg.1 ENST00000427796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100198610.hg.1 NR_038280.1 Refseq
          Human Genome U95 Set 32048_at - -
          Human Genome U95 Set 32048_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031566 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031566 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 129526 - -
          ABI Human Genome Survey Microarray Version 2.0 214258 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0