Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00297288_s1
          See other NAT8L GE Assays ›
          Gene Symbol:
          NAT8L
          Gene Name:
          N-acetyltransferase 8 like
          Gene Aliases:
          CML3, NACED, NAT8-LIKE
          Chromosome Location:
          Chr.4: 2059512 - 2069089 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00297288_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NAT8L
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178557.3 NP_848652.2 3 3850 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NAT8L

          Gene Ontology Categories:

          Process(es)

          aspartate N-acetyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23038651 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23038643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892081 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23038650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892092 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23038657 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23038654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892102 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892071 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892106 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892074 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23038634 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23038637 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15534402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02892070 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 620424 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3670415 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P91991 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P91991 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 3652 - -
          Agilent Human Genome CGH Microarray 44B A_14_P123072 - -
          Agilent Human Genome CGH Microarray 44A A_14_P123072 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 50049 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 5511 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00908164 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00908168 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3670415 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P91991 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3670415 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P91991 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892084 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23038633 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23038624 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23038599 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15534402 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15534411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23038633 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892084 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23038624 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23038599 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15534402 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15534411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02892077 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P91991 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P123072 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892082 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892070 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892081 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892077 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892073 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892076 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892075 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892084 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892071 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892079 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892085 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02892074 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534393 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534391 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534399 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534424 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892091 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892076 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892094 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534401 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892095 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892093 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534422 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534385 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534420 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534390 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534395 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534389 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534406 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892077 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534405 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892092 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534386 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534392 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892075 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534394 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534403 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534397 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534423 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534404 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534396 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534407 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892090 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534400 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534398 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534387 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534402 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534408 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15534388 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02892074 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P123072 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P123072 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P91991 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 18353 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 20868 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00908164 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892089 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892082 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892110 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892079 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892117 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892100 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534407 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892111 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892084 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892113 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534395 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892101 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892073 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892081 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892103 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892109 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892087 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892106 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534402 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892116 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534414 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892108 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892090 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892096 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892092 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892095 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892077 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534393 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534435 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892105 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534389 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892094 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534386 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534411 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892070 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534401 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02892115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15534427 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_178557_at - -
          Affymetrix Human Gene 1.0 ST Array 8093587 - -
          Affymetrix Human Gene 1.0 ST Array 8093584 - -
          Affymetrix Human Gene 1.0 ST Array 8093586 - -
          Affymetrix Human Gene 1.0 ST Array 8093585 - -
          Affymetrix Human Gene 1.0 ST Array 8093582 - -
          Affymetrix Human Gene 1.0 ST Array 8093581 - -
          Affymetrix Human Gene 1.0 ST Array 8093583 - -
          Affymetrix Human Gene 1.1 ST Array 8093578 - -
          Affymetrix HT HG-U133+ PM Array Plate 235316_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 228880_PM_at - -
          Affymetrix Human Genome U219 Array 11733272_PM_x_at - -
          Affymetrix Human Genome U219 Array 11733272_x_at - -
          Affymetrix Human Genome U219 Array 11733269_PM_at - -
          Affymetrix Human Genome U219 Array 11733271_at - -
          Affymetrix Human Genome U219 Array 11733271_PM_at - -
          Affymetrix Human Genome U219 Array 11733270_at - -
          Affymetrix Human Genome U219 Array 11733269_at - -
          Affymetrix Human Genome U219 Array 11733270_PM_at - -
          Human Genome U133 Plus 2.0 Array 235316_at - -
          Human Genome U133 Plus 2.0 Array 228880_PM_at - -
          Human Genome U133 Plus 2.0 Array 228880_at - -
          Human Genome U133 Plus 2.0 Array 235316_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_178557_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235316_at1 - -
          Human Genome U95 Set 74943_PM_at - -
          Human Genome U95 Set 74943_at - -
          Human Genome U133 Set 228880_at - -
          Human Genome U133 Set 228880_PM_at - -
          Human Genome U133 Set 235316_PM_at - -
          Human Genome U133 Set 235316_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025171 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025171 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3237830 AK094797.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3237830 NM_178557.3 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1672437 - -
          HumanHT-12 V4.0 expression beadchip ILMN_3237830 AK094797.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3237830 NM_178557.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3237830 AK094797.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3237830 NM_178557.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3237830 AK094797.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3237830 NM_178557.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31341708-S AK094797.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31341708-S NM_178557.3 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_20605-S - -
          Illumina human-6 v1.0 expression beadchip 0006200164 - -
          Illumina humanRef-8 v1.0 expression beadchip 0006200164 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1672437 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1672437 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 4390451 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_20605 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1672437 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0