Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000429014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000608965.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000610149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013444.hg.1 |
ENST00000621535.1 |
Ensembl |