Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001165.hg.1 |
ENST00000355072.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001165.hg.1 |
NM_002111.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001165.hg.1 |
TC04000044.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0400145622.hg.1 |
ENST00000355072.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145622.hg.1 |
ENST00000510626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145622.hg.1 |
NM_002111.8 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145622.hg.1 |
TC0400006591.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400145622.hg.1 |
NM_002111.8 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145622.hg.1 |
ENST00000355072.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145622.hg.1 |
ENST00000510626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145621.hg.1 |
ENST00000355072.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145621.hg.1 |
ENST00000510626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145621.hg.1 |
NM_002111.8 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145621.hg.1 |
TC0400006591.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400145621.hg.1 |
ENST00000355072.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145621.hg.1 |
ENST00000510626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145621.hg.1 |
NM_002111.8 |
Refseq |