Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00356153_g1
          See other CHUK GE Assays ›
          Gene Symbol:
          CHUK
          Gene Name:
          conserved helix-loop-helix ubiquitous kinase
          Gene Aliases:
          IKBKA, IKK-alpha, IKK1, IKKA, NFKBIKA, TCF16
          Chromosome Location:
          Chr.10: 100188298 - 100229610 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00356153_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CHUK
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001278.4 NP_001269.3 6 - 7 639 97
          NM_001320928.1 NP_001307857.1 6 - 7 639 97
          XM_017015611.1 XP_016871100.1 6 - 7 639 97
          XM_017015612.1 XP_016871101.1 6 - 7 639 97
          GenBank mRNA AB209028.1 - 6 - 7 593 97
          AF009225.1 - 6 - 7 596 97
          AF012890.1 - 6 - 7 561 97
          AF080157.1 - 6 - 7 656 97
          BC092514.1 - 6 - 7 577 3063613 97
          U22512.1 - 5 - 6 469 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000012585.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10002931.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CHUK

          Gene Ontology Categories:

          Function(s) Process(es)

          stimulatory C-type lectin receptor signaling pathway
          skeletal muscle contraction
          protein phosphorylation
          inflammatory response
          immune response
          I-kappaB kinase/NF-kappaB signaling
          I-kappaB phosphorylation
          Rho protein signal transduction
          response to xenobiotic stimulus
          response to virus
          response to toxic substance
          anatomical structure morphogenesis
          response to acetate
          regulation of tumor necrosis factor-mediated signaling pathway
          response to lipopolysaccharide
          response to hydroperoxide
          TRIF-dependent toll-like receptor signaling pathway
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to amino acid
          innate immune response
          positive regulation of transcription by RNA polymerase II
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          striated muscle cell differentiation
          stress-activated MAPK cascade
          nucleotide-binding oligomerization domain containing signaling pathway
          cellular response to tumor necrosis factor
          protein kinase activity
          protein binding
          ATP binding
          IkappaB kinase activity
          protein homodimerization activity
          protein heterodimerization activity
          scaffold protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10020492.hg.1 ENST00000370397.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10020492.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10020492.hg.1 TC10002931.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000585551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000585551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000585551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000585551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10020491.hg.1 ENST00000370397.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10020491.hg.1 NM_001278.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10020491.hg.1 TC10002931.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000187793.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000187793.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000187793.hg.1 TC1000012585.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000187793.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000187793.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000187795.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000187795.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000187795.hg.1 TC1000012585.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000187795.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000187795.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000370397.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000585551.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000585551.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000585551.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000585551.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000588656.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 ENST00000590930.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012585.hg.1 NM_001278.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0