Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00357612_g1
          See other ATP1B1 GE Assays ›
          Gene Symbol:
          ATP1B1
          Gene Name:
          ATPase Na+/K+ transporting subunit beta 1
          Gene Aliases:
          ATP1B
          Chromosome Location:
          Chr.1: 169106709 - 169132722 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00357612_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATP1B1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001677.3 NP_001668.1 2 - 3 349 111
          GenBank mRNA AK290181.1 - 2 - 3 362 111
          AK296059.1 - 2 - 3 237 111
          AK316043.1 - 2 - 3 294 111
          AY545986.1 - 2 - 3 228 111
          BC000006.2 - 2 - 3 314 3506311 111
          BQ934151.1 - 3 - 4 553 111
          BT009787.1 - 2 - 3 228 111
          CR407616.1 - 2 - 3 228 111
          U16799.1 - 2 - 3 273 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010543.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001473.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATP1B1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of defense response to virus by host
          cellular calcium ion homeostasis
          cellular sodium ion homeostasis
          cell adhesion
          establishment or maintenance of transmembrane electrochemical gradient
          regulation of gene expression
          regulation of cardiac muscle contraction by calcium ion signaling
          cellular potassium ion homeostasis
          positive regulation of ATPase activity
          ion transmembrane transport
          sodium ion export across plasma membrane
          protein transport into plasma membrane raft
          ATP metabolic process
          protein stabilization
          leukocyte migration
          transmembrane transport
          relaxation of cardiac muscle
          cardiac muscle contraction
          protein localization to plasma membrane
          membrane repolarization
          membrane repolarization during cardiac muscle cell action potential
          cell communication by electrical coupling involved in cardiac conduction
          positive regulation of mitophagy in response to mitochondrial depolarization
          xenophagy
          positive regulation of potassium ion transmembrane transporter activity
          positive regulation of sodium ion export across plasma membrane
          positive regulation of calcium:sodium antiporter activity
          positive regulation of potassium ion import across plasma membrane
          regulation of cardiac conduction
          potassium ion import across plasma membrane
          ATPase activator activity
          P-type sodium:potassium-exchanging transporter activity
          protein binding
          ATP binding
          protein C-terminus binding
          ATP hydrolysis activity
          MHC class II protein complex binding
          potassium ion binding
          sodium ion binding
          ATPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01022845.hg.1 ENST00000367813.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022845.hg.1 ENST00000367815.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022845.hg.1 ENST00000367816.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022845.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022845.hg.1 ENST00000499679.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022845.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01022845.hg.1 TC01001473.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01022844.hg.1 ENST00000367813.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022844.hg.1 ENST00000367815.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022844.hg.1 ENST00000367816.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022844.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022844.hg.1 ENST00000499679.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01022844.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01022844.hg.1 TC01001473.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010543.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100177597.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177597.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177597.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177597.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100177597.hg.1 TC0100010543.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100177597.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177597.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177597.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177597.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100177596.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177596.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177596.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177596.hg.1 NM_001677.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100177596.hg.1 TC0100010543.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100177596.hg.1 ENST00000367815.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177596.hg.1 ENST00000367816.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177596.hg.1 ENST00000494797.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100177596.hg.1 NM_001677.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0