Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027610.hg.1 |
ENST00000003302.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027610.hg.1 |
ENST00000260188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027610.hg.1 |
ENST00000544967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027610.hg.1 |
ENST00000545540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027610.hg.1 |
NM_020886.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027610.hg.1 |
TC11002307.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
ENST00000003302.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
ENST00000540438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
ENST00000544967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
ENST00000545540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
NM_001301029.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
NM_020886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
TC1100012358.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
ENST00000544967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
ENST00000003302.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
ENST00000540438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
ENST00000545540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
NM_001301029.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194958.hg.1 |
NM_020886.3 |
Refseq |