Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTTNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033427.2 | NP_219499.1 | 12 - 13 | 3445 | 65 | |
XM_005250634.4 | XP_005250691.1 | 12 - 13 | 3580 | 65 | ||
XM_005250635.3 | XP_005250692.1 | 12 - 13 | 3214 | 65 | ||
XM_011516615.2 | XP_011514917.1 | 12 - 13 | 5748 | 65 | ||
XM_011516616.2 | XP_011514918.1 | 12 - 13 | 3377 | 65 | ||
XM_017012706.1 | XP_016868195.1 | 12 - 13 | 3377 | 65 | ||
XM_017012708.1 | XP_016868197.1 | 9 - 10 | 2959 | 65 | ||
GenBank mRNA | AB051545.1 | - | 12 - 13 | 3351 | 65 | |
AF377960.1 | - | 12 - 13 | 3445 | 65 | ||
BC106000.1 | - | 12 - 13 | 3412 | 30528261 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |