Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005232.hg.1 |
ENST00000317593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005232.hg.1 |
ENST00000513010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005232.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005232.hg.1 |
TC05000373.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005231.hg.1 |
ENST00000513010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005231.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005231.hg.1 |
TC05000373.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005233.hg.1 |
ENST00000317593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005233.hg.1 |
ENST00000513010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005233.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005233.hg.1 |
TC05000373.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000613039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000613039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000613039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
ENST00000613039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153620.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153620.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153620.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153620.hg.1 |
TC0500007836.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500153620.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153620.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153620.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153619.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153619.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153619.hg.1 |
TC0500007836.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500153619.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153619.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153621.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153621.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153621.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153621.hg.1 |
TC0500007836.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500153621.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153621.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153621.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000317593.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000513010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000613039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000613039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000613039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
ENST00000613039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007836.hg.1 |
NM_130772.3 |
Refseq |