Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
ENST00000461027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
ENST00000461027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
ENST00000461027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
ENST00000367660.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
ENST00000461027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
ENST00000461027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
ENST00000461027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009626.hg.1 |
NM_021243.2 |
Refseq |