Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00386700_m1
          See other PLA2G1B GE Assays ›
          Gene Symbol:
          PLA2G1B
          Gene Name:
          phospholipase A2 group IB
          Gene Aliases:
          PLA2, PLA2A, PPLA2
          Chromosome Location:
          Chr.12: 120322111 - 120327789 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00386700_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PLA2G1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000928.2 NP_000919.1 2 - 3 237 126
          GenBank mRNA AA844927.1 - 2 - 3 126
          AK311830.1 - 2 - 3 222 126
          BC106725.1 - 2 - 3 237 40014551 126
          BC106726.1 - 2 - 3 237 40014552 126
          BE969737.1 - 208 126
          CA867923.1 - 1 - 2 80 126
          M21054.1 - 2 - 3 238 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200012158.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12002042.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PLA2G1B

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          neutrophil mediated immunity
          fatty acid biosynthetic process
          phosphatidic acid biosynthetic process
          actin filament organization
          signal transduction
          positive regulation of calcium ion transport into cytosol
          lipid catabolic process
          leukotriene biosynthetic process
          antibacterial humoral response
          neutrophil chemotaxis
          activation of phospholipase A2 activity
          interleukin-8 production
          cellular response to insulin stimulus
          intracellular signal transduction
          phosphatidylglycerol acyl-chain remodeling
          phosphatidylinositol acyl-chain remodeling
          phosphatidylserine acyl-chain remodeling
          phosphatidylcholine acyl-chain remodeling
          phosphatidylethanolamine acyl-chain remodeling
          positive regulation of DNA replication
          positive regulation of transcription by RNA polymerase II
          phosphatidylcholine metabolic process
          positive regulation of fibroblast proliferation
          arachidonic acid secretion
          positive regulation of protein secretion
          positive regulation of immune response
          defense response to Gram-positive bacterium
          positive regulation of NF-kappaB transcription factor activity
          glucose transmembrane transport
          phospholipase A2 activity
          signaling receptor binding
          calcium ion binding
          bile acid binding
          calcium-dependent phospholipase A2 activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12026828.hg.1 ENST00000308366.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12026828.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12026828.hg.1 NM_000928.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12026828.hg.1 TC12002042.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000423423.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012158.hg.1 NM_000928.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200194807.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200194807.hg.1 ENST00000549767.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200194807.hg.1 NM_000928.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200194807.hg.1 TC1200012158.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200194807.hg.1 NM_000928.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200194807.hg.1 ENST00000308366.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200194807.hg.1 ENST00000549767.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0