Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00391179_m1
          See other CLASP2 GE Assays ›
          Gene Symbol:
          CLASP2
          Gene Name:
          cytoplasmic linker associated protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 33496245 - 33718356 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00391179_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CLASP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001207044.1 NP_001193973.1 14 - 15 1724 67
          NM_015097.2 NP_055912.2 20 - 21 2185 67
          XM_006713040.1 XP_006713103.1 20 - 21 2328 67
          XM_006713041.1 XP_006713104.1 20 - 21 2325 67
          XM_006713042.1 XP_006713105.1 20 - 21 2328 67
          XM_006713048.1 XP_006713111.1 20 - 21 2328 67
          XM_006713049.1 XP_006713112.1 20 - 21 2325 67
          XM_006713050.1 XP_006713113.1 20 - 21 2328 67
          XM_006713052.1 XP_006713115.1 20 - 21 2325 67
          XM_006713053.1 XP_006713116.1 20 - 21 2328 67
          XM_017005947.1 XP_016861436.1 20 - 21 2325 67
          XM_017005948.1 XP_016861437.1 20 - 21 2325 67
          XM_017005950.1 XP_016861439.1 20 - 21 2328 67
          XM_017005951.1 XP_016861440.1 20 - 21 2325 67
          XM_017005952.1 XP_016861441.1 20 - 21 2328 67
          XM_017005953.1 XP_016861442.1 20 - 21 2325 67
          XM_017005954.1 XP_016861443.1 20 - 21 2325 67
          XM_017005955.1 XP_016861444.1 20 - 21 2328 67
          XM_017005956.1 XP_016861445.1 20 - 21 2328 67
          XM_017005957.1 XP_016861446.1 20 - 21 2328 67
          XM_017005958.1 XP_016861447.1 20 - 21 2328 67
          XM_017005959.1 XP_016861448.1 20 - 21 2325 67
          XM_017005962.1 XP_016861451.1 20 - 21 2325 67
          XM_017005963.1 XP_016861452.1 20 - 21 2328 67
          XM_017005964.1 XP_016861453.1 20 - 21 2325 67
          XM_017005968.1 XP_016861457.1 14 - 15 1468 67
          XM_017005969.1 XP_016861458.1 14 - 15 1478 67
          XM_017005970.1 XP_016861459.1 14 - 15 1727 67
          XM_017005971.1 XP_016861460.1 14 - 15 1478 67
          XM_017005972.1 XP_016861461.1 14 - 15 2055 67
          XM_017005973.1 XP_016861462.1 14 - 15 1724 67
          XM_017005974.1 XP_016861463.1 14 - 15 1724 67
          XM_017005975.1 XP_016861464.1 14 - 15 1468 67
          XM_017005977.1 XP_016861466.1 14 - 15 1727 67
          XM_017005978.1 XP_016861467.1 14 - 15 1468 67
          XM_017005979.1 XP_016861468.1 14 - 15 1478 67
          XM_017005980.1 XP_016861469.1 14 - 15 2055 67
          XM_017005981.1 XP_016861470.1 14 - 15 1468 67
          XM_017005982.1 XP_016861471.1 14 - 15 1478 67
          XM_017005983.1 XP_016861472.1 14 - 15 1727 67
          XM_017005984.1 XP_016861473.1 14 - 15 1724 67
          XM_017005985.1 XP_016861474.1 14 - 15 2055 67
          GenBank mRNA AB014527.2 - 14 - 15 1363 67
          AK091451.1 - 20 - 21 2325 67
          AK297322.1 - 13 - 14 1387 67
          BC140778.1 - 20 - 21 2021 9021646 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300010676.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001277.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CLASP2

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          vesicle targeting
          microtubule nucleation
          negative regulation of microtubule depolymerization
          Golgi organization
          mitotic spindle organization
          sister chromatid cohesion
          establishment or maintenance of cell polarity
          exit from mitosis
          regulation of gastrulation
          positive regulation of epithelial cell migration
          regulation of epithelial to mesenchymal transition
          microtubule organizing center organization
          regulation of microtubule polymerization or depolymerization
          regulation of microtubule-based process
          microtubule anchoring
          positive regulation of exocytosis
          cell division
          negative regulation of stress fiber assembly
          negative regulation of focal adhesion assembly
          protein localization to plasma membrane
          positive regulation of extracellular matrix disassembly
          negative regulation of wound healing, spreading of epidermal cells
          positive regulation of basement membrane assembly involved in embryonic body morphogenesis
          dystroglycan binding
          protein binding
          microtubule binding
          microtubule plus-end binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 ENST00000307312.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 ENST00000359576.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 ENST00000399362.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 ENST00000461133.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 ENST00000468888.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 ENST00000480013.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 ENST00000539981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 NM_001207044.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 NM_015097.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03021927.hg.1 TC03001277.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000359576.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000399362.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000461133.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000468888.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000480013.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 NM_001207044.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 NM_015097.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 TC0300010676.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000359576.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000399362.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000461133.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000468888.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 ENST00000480013.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 NM_001207044.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300177732.hg.1 NM_015097.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline