Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00391673_m1
          See other DST GE Assays ›
          Gene Symbol:
          DST
          Gene Name:
          dystonin
          Gene Aliases:
          BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
          Chromosome Location:
          Chr.6: 56457987 - 56954671 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00391673_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DST
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001144769.2 NP_001138241.1 66 - 67 12263 92
          NM_001144770.1 NP_001138242.1 64 - 65 11934 92
          NM_015548.4 NP_056363.2 53 - 54 10818 92
          NM_183380.3 NP_899236.1 63 - 64 12208 92
          XM_005249310.3 XP_005249367.1 72 - 73 18615 92
          XM_005249315.3 XP_005249372.1 71 - 72 18562 92
          XM_005249316.2 XP_005249373.1 64 - 65 18838 92
          XM_005249318.2 XP_005249375.1 70 - 71 18276 92
          XM_005249319.2 XP_005249376.1 69 - 70 18177 92
          XM_005249320.4 XP_005249377.1 69 - 70 18233 92
          XM_005249322.4 XP_005249379.1 69 - 70 18128 92
          XM_005249323.3 XP_005249380.1 68 - 69 18461 92
          XM_005249324.2 XP_005249381.1 58 - 59 17073 92
          XM_011514824.2 XP_011513126.1 71 - 72 18526 92
          XM_011514825.2 XP_011513127.1 66 - 67 12264 92
          XM_011514826.2 XP_011513128.1 67 - 68 18134 92
          XM_017011205.1 XP_016866694.1 72 - 73 18625 92
          XM_017011206.1 XP_016866695.1 72 - 73 18625 92
          XM_017011207.1 XP_016866696.1 72 - 73 18562 92
          XM_017011208.1 XP_016866697.1 72 - 73 18625 92
          XM_017011209.1 XP_016866698.1 72 - 73 18625 92
          XM_017011210.1 XP_016866699.1 72 - 73 18625 92
          XM_017011211.1 XP_016866700.1 72 - 73 18625 92
          XM_017011212.1 XP_016866701.1 71 - 72 18391 92
          XM_017011213.1 XP_016866702.1 71 - 72 18298 92
          XM_017011214.1 XP_016866703.1 71 - 72 18298 92
          XM_017011215.1 XP_016866704.1 71 - 72 18298 92
          XM_017011216.1 XP_016866705.1 71 - 72 18298 92
          XM_017011217.1 XP_016866706.1 69 - 70 18206 92
          XM_017011218.1 XP_016866707.1 34 - 35 14234 92
          XM_017011219.1 XP_016866708.1 67 - 68 12363 92
          XM_017011220.1 XP_016866709.1 66 - 67 12306 92
          XM_017011221.1 XP_016866710.1 66 - 67 12129 92
          XM_017011222.1 XP_016866711.1 59 - 60 12522 92
          XM_017011223.1 XP_016866712.1 66 - 67 12036 92
          XM_017011224.1 XP_016866713.1 62 - 63 11876 92
          GenBank mRNA AB018271.2 - 1 - 2 108 92
          AF400226.1 - 53 - 54 92
          AK055189.1 - 13 - 14 3548 92
          AK096713.1 - 9 - 10 1547 92
          AK128632.1 - 12 - 13 2352 92

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DST

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          cell adhesion
          integrin-mediated signaling pathway
          response to wounding
          maintenance of cell polarity
          intermediate filament cytoskeleton organization
          cell motility
          actin binding
          integrin binding
          calcium ion binding
          protein binding
          protein C-terminus binding
          microtubule plus-end binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0