Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00391697_g1
          See other DST GE Assays ›
          Gene Symbol:
          DST
          Gene Name:
          dystonin
          Gene Aliases:
          BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
          Chromosome Location:
          Chr.6: 56457987 - 56954671 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00391697_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DST
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001144769.2 NP_001138241.1 90 - 91 16161 72
          NM_001144770.1 NP_001138242.1 88 - 89 15832 72
          NM_015548.4 NP_056363.2 77 - 78 14716 72
          NM_183380.3 NP_899236.1 87 - 88 16106 72
          XM_005249310.3 XP_005249367.1 96 - 97 22513 72
          XM_005249315.3 XP_005249372.1 95 - 96 22460 72
          XM_005249316.2 XP_005249373.1 88 - 89 22736 72
          XM_005249318.2 XP_005249375.1 94 - 95 22174 72
          XM_005249319.2 XP_005249376.1 93 - 94 22075 72
          XM_005249320.4 XP_005249377.1 93 - 94 22131 72
          XM_005249322.4 XP_005249379.1 93 - 94 22026 72
          XM_005249323.3 XP_005249380.1 92 - 93 22359 72
          XM_005249324.2 XP_005249381.1 82 - 83 20971 72
          XM_011514824.2 XP_011513126.1 95 - 96 22424 72
          XM_011514825.2 XP_011513127.1 90 - 91 16162 72
          XM_011514826.2 XP_011513128.1 91 - 92 22032 72
          XM_017011205.1 XP_016866694.1 96 - 97 22523 72
          XM_017011206.1 XP_016866695.1 96 - 97 22523 72
          XM_017011207.1 XP_016866696.1 96 - 97 22460 72
          XM_017011208.1 XP_016866697.1 96 - 97 22523 72
          XM_017011209.1 XP_016866698.1 96 - 97 22523 72
          XM_017011210.1 XP_016866699.1 96 - 97 22523 72
          XM_017011211.1 XP_016866700.1 96 - 97 22523 72
          XM_017011212.1 XP_016866701.1 95 - 96 22289 72
          XM_017011213.1 XP_016866702.1 95 - 96 22196 72
          XM_017011214.1 XP_016866703.1 95 - 96 22196 72
          XM_017011215.1 XP_016866704.1 95 - 96 22196 72
          XM_017011216.1 XP_016866705.1 95 - 96 22196 72
          XM_017011217.1 XP_016866706.1 93 - 94 22104 72
          XM_017011218.1 XP_016866707.1 58 - 59 18132 72
          XM_017011219.1 XP_016866708.1 91 - 92 16261 72
          XM_017011220.1 XP_016866709.1 90 - 91 16204 72
          XM_017011221.1 XP_016866710.1 90 - 91 16027 72
          XM_017011222.1 XP_016866711.1 83 - 84 16420 72
          XM_017011223.1 XP_016866712.1 90 - 91 15934 72
          XM_017011224.1 XP_016866713.1 86 - 87 15774 72
          GenBank mRNA AB018271.2 - 25 - 26 4006 72
          AF400226.1 - 77 - 78 14608 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600012123.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001828.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DST

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          cell adhesion
          integrin-mediated signaling pathway
          response to wounding
          maintenance of cell polarity
          intermediate filament cytoskeleton organization
          cell motility
          actin binding
          integrin binding
          calcium ion binding
          protein binding
          protein C-terminus binding
          microtubule plus-end binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 ENST00000244364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 ENST00000312431.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 ENST00000361203.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 ENST00000370754.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 ENST00000370769.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 ENST00000370788.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 ENST00000421834.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 ENST00000446842.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 NM_015548.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06022136.hg.1 TC06001828.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000244364.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000312431.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000361203.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000370754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000370788.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000421834.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000523292.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 NM_001144769.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 NM_001144770.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 NM_015548.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 NM_183380.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 TC0600012123.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000244364.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000312431.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000361203.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000370754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000370788.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000421834.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 ENST00000523292.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 NM_001144769.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 NM_001144770.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 NM_015548.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600183513.hg.1 NM_183380.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0