Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00400907_m1
          See other SMLR1 GE Assays ›
          Gene Symbol:
          SMLR1
          Gene Name:
          small leucine rich protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 130827405 - 130837135 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00400907_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SMLR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195597.1 NP_001182526.1 1 - 2 248 75
          GenBank mRNA AK091900.1 - 1 - 2 247 75
          BC101422.1 - 1 - 2 247 75
          BC101423.1 - 1 - 2 247 75
          BC101424.1 - 1 - 2 247 75
          BC101425.1 - 1 - 2 247 75
          BC113026.1 - 1 - 2 247 75
          BC113027.1 - 1 - 2 248 75
          BX095531.1 - 1 - 2 186 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600009445.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000981.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276135 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15862041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05075776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05075780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05075760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15862035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15862040 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276128 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05075778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05075751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05075762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15862037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276120 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276116 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25276138 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 35819 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 40255 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 757026 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P463929 - -
          Agilent Human Genome CGH Microarray 44B A_14_P201056 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P463929 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P463929 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P201056 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P201056 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 91653 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 90394 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17722902 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15845172 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P201056 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37799009 - -
          Agilent Human 1B Microarray G4111A A_32_P91669 - -
          Agilent Human 1B Microarray G4111A A_32_P431352 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P201056 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600009445.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009445.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009445.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009445.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009445.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009445.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011535.hg.1 ENST00000541421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011535.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011535.hg.1 TC06000981.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06011536.hg.1 ENST00000541421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011536.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011536.hg.1 TC06000981.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600164759.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600164759.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600164759.hg.1 TC0600009445.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600164759.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600164759.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600164760.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600164760.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600164760.hg.1 TC0600009445.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600164760.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600164760.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Clariom D Assay TC0600009445.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009445.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009445.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009445.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Clariom D Assay TC0600009445.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Clariom D Assay TC0600009445.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600164758.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600164758.hg.1 NM_001195597.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600164758.hg.1 TC0600009445.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600164758.hg.1 ENST00000541421.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600164758.hg.1 NM_001195597.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 196325 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline