Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00404172_m1
          See other RPS7P5 GE Assays ›
          Gene Symbol:
          RPS7P5
          Gene Name:
          ribosomal protein S7 pseudogene 5
          Gene Aliases:
          RPS7_2_183
          Chromosome Location:
          Chr.1: 240007524 - 240013260 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00404172_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RPS7P5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036695.1 1 - 2 314 64
          GenBank mRNA AK098605.1 - 1 - 2 314 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100012148.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001965.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969624 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969637 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898568 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969632 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969679 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898537 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898635 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15197244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969651 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898562 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15197234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00898550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20969585 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P15765 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P15765 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 33722 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 51367 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P15765 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P15765 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P15765 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10571182 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15504250 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10571421 - -
          Agilent Human 1B Microarray G4111A A_32_P763622 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01030477.hg.1 NR_036695.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01030477.hg.1 TC01001965.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01030478.hg.1 NR_036695.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01030478.hg.1 TC01001965.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100012148.hg.1 ENST00000441482.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012148.hg.1 NR_036695.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012148.hg.1 NR_036695.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012148.hg.1 NR_036695.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012148.hg.1 NR_036695.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400010476.hg.1 ENST00000513977.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400010476.hg.1 ENST00000513977.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006527.hg.1 ENST00000596308.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006527.hg.1 ENST00000596308.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006527.hg.1 ENST00000596308.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006527.hg.1 ENST00000597891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006527.hg.1 ENST00000597891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000417099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000417099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000418950.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000418950.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000424604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000424604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000424604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000577672.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000577672.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000577672.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006531.hg.1 ENST00000577672.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100189451.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100189451.hg.1 TC0100012148.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100189451.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay TC0400010476.hg.1 ENST00000513977.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400010476.hg.1 ENST00000513977.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100189454.hg.1 ENST00000441482.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100189454.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100189454.hg.1 TC0100012148.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100189454.hg.1 ENST00000441482.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100189454.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100189455.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100189455.hg.1 TC0100012148.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100189455.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000417099.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000417099.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000418950.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000418950.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000424604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000424604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000424604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000577672.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000577672.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000577672.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006531.hg.1 ENST00000577672.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100189452.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100189452.hg.1 TC0100012148.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100189452.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012148.hg.1 ENST00000441482.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012148.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012148.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012148.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012148.hg.1 NR_036695.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006527.hg.1 ENST00000596308.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006527.hg.1 ENST00000596308.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006527.hg.1 ENST00000596308.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006527.hg.1 ENST00000597891.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006527.hg.1 ENST00000597891.1 Ensembl
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042235 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042235 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 228045 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0