Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001376.4 | NP_001367.2 | 41 - 42 | 8509 | 101 | |
GenBank mRNA | AB002323.3 | - | 41 - 42 | 8381 | 101 | |
AB290157.1 | - | 41 - 42 | 8509 | 101 | ||
AK023747.1 | - | 7 - 8 | 1623 | 101 | ||
AK095415.1 | - | 11 - 12 | 2122 | 101 | ||
AL833600.1 | - | 13 - 14 | 2507 | 101 | ||
BX648670.1 | - | 6 - 7 | 1111 | 101 |
Set Membership: |
![]() ![]() ![]() ![]() |