Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00415851_g1
          See other CDC20 GE Assays ›
          Gene Symbol:
          CDC20
          Gene Name:
          cell division cycle 20
          Gene Aliases:
          CDC20A, bA276H19.3, p55CDC
          Chromosome Location:
          Chr.1: 43358955 - 43363203 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00415851_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDC20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001255.2 NP_001246.2 1 - 2 50 108
          GenBank mRNA AK302539.1 - 1 - 2 49 108
          AK303844.1 - 1 - 2 77 108
          BC001088.2 - 1 - 2 50 2964686 108
          BC010044.2 - 1 - 2 41 3634656 108
          BC012827.2 - 1 - 2 50 2964686 108
          U05340.1 - 1 - 2 59 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100008057.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000545.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK302539.1,AK303844.1,BC001088.2,BC010044.2,BC012827.2,NM_001255.2,U05340.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CDC20

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          sister chromatid cohesion
          mitotic nuclear division
          positive regulation of cell proliferation
          anaphase-promoting complex-dependent catabolic process
          positive regulation of synaptic plasticity
          regulation of meiotic nuclear division
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of dendrite development
          cell division
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of synapse maturation
          positive regulation of ubiquitin protein ligase activity
          protein binding
          protein C-terminus binding
          anaphase-promoting complex binding
          enzyme binding
          histone deacetylase binding
          ubiquitin-protein transferase activator activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01008662.hg.1 TC01000545.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01008662.hg.1 ENST00000310955.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008662.hg.1 ENST00000437896.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008662.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000482046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000482046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000482046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 ENST00000482046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01008663.hg.1 TC01000545.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01008663.hg.1 ENST00000310955.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008663.hg.1 ENST00000437896.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008663.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01008664.hg.1 TC01000545.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01008664.hg.1 ENST00000310955.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008664.hg.1 ENST00000437896.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008664.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007800.hg.1 ENST00000488829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008665.hg.1 TC01000545.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01008665.hg.1 ENST00000310955.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008665.hg.1 ENST00000437896.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008665.hg.1 NM_001255.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01008669.hg.1 TC01000545.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01008669.hg.1 ENST00000310955.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008669.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008669.hg.1 ENST00000437896.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008669.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000478882.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000482046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000482046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000482046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 ENST00000482046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008057.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100156891.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156891.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156891.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100156891.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156891.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156891.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100156891.hg.1 TC0100008057.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100156892.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156892.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156892.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100156892.hg.1 ENST00000310955.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156892.hg.1 ENST00000372462.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156892.hg.1 NM_001255.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100156892.hg.1 TC0100008057.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0900007800.hg.1 ENST00000488829.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 106187 - -
          ABI Human Genome Survey Microarray Version 2.0 161150 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0