Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006798.hg.1 |
ENST00000433810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006798.hg.1 |
ENST00000433810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006798.hg.1 |
ENST00000433810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006798.hg.1 |
NR_125783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006798.hg.1 |
NR_125783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006798.hg.1 |
NR_125783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146605.hg.1 |
ENST00000433810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146605.hg.1 |
NR_125783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146605.hg.1 |
TC0200006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200146605.hg.1 |
NR_125783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146605.hg.1 |
ENST00000433810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006798.hg.1 |
ENST00000433810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006798.hg.1 |
ENST00000433810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006798.hg.1 |
ENST00000433810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006798.hg.1 |
NR_125783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006798.hg.1 |
NR_125783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006798.hg.1 |
NR_125783.1 |
Refseq |