Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000380437.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000434457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000494124.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000602442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000602925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
NM_017925.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
NR_073201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
TC0900006758.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000380437.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000434457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000494124.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000602442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
ENST00000602925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
NM_017925.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900146106.hg.1 |
NR_073201.3 |
Refseq |