Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00417186_m1
          See other GS1-279B7.1 GE Assays ›
          Gene Symbol:
          GS1-279B7.1
          Gene Name:
          microtubule associated protein 1 light chain 3 beta pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 185323846 - 185335039 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00417186_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GS1-279B7.1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038424.1 3 - 4 482 124
          GenBank mRNA AF508905.1 - 3 - 4 450 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100016695.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003630.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656451 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716276 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15151704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15151708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656425 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716290 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656433 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716282 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15151701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656469 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15151698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15151700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00656472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15151702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20716239 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 50338 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 898552 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 123636 AK311879.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 123636 BC018386.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 123636 NM_001866.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 123636 Z14244.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 350989 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P35389532 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35389532 - -
          Agilent Human 1B Microarray G4111A A_32_P440943 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P35389532 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01055842.hg.1 ENST00000446901.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055842.hg.1 ENST00000457764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055842.hg.1 NR_038424.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01055842.hg.1 TC01003630.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01055841.hg.1 ENST00000446901.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055841.hg.1 NR_038424.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01055841.hg.1 TC01003630.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01055844.hg.1 ENST00000423844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055844.hg.1 ENST00000446901.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055844.hg.1 ENST00000457764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055844.hg.1 NR_038424.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01055844.hg.1 TC01003630.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226639.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226639.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226639.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226639.hg.1 TC0100016695.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100226639.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226639.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226639.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016695.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226640.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226640.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226640.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226640.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226640.hg.1 TC0100016695.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100226640.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226640.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226640.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226640.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 NR_038424.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 TC0100016695.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 ENST00000562817.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 ENST00000563533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 ENST00000565520.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226643.hg.1 NR_038424.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008914 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008914 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 162183 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0