Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900202880.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202880.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202880.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900202880.hg.1 |
TC1900011809.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900202880.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900202880.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202880.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202879.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202879.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202879.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900202879.hg.1 |
TC1900011809.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900202879.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202879.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202879.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900202881.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202881.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900202881.hg.1 |
TC1900011809.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900202881.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202881.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900202878.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202878.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900202878.hg.1 |
TC1900011809.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900202878.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202878.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000595954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
ENST00000597946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011809.hg.1 |
NR_110744.1 |
Refseq |