Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00419420_m1
          See other UBAC2-AS1 GE Assays ›
          Gene Symbol:
          UBAC2-AS1
          Gene Name:
          UBAC2 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 99196374 - 99200757 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00419420_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UBAC2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036531.1 1 - 2 393 81
          GenBank mRNA AK097488.1 - 1 - 2 398 81
          DB032212.1 - 1 - 2 394 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300009591.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000826.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09073994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29523173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09073993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29523176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29523175 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29523178 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 288606 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 930068 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P115122 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02846406 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02846406 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19928538 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02846406 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1557038_PM_s_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1557037_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR13009317.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009317.hg.1 NR_036532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009317.hg.1 TC13000826.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR13009319.hg.1 ENST00000426037.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009319.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009319.hg.1 TC13000826.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 TC1300009591.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164183.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 TC1300009591.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164185.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164188.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164188.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164188.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164188.hg.1 TC1300009591.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300164188.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164188.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164188.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 TC1300009591.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164184.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009591.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 NR_036532.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 TC1300009591.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 ENST00000426037.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 ENST00000445737.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 NR_036531.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164186.hg.1 NR_036532.1 Refseq
          Human Genome U133 Plus 2.0 Array 1557037_a_at - -
          Human Genome U133 Plus 2.0 Array 1557038_s_at - -
          Human Genome U133 Plus 2.0 Array 1557038_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 1557037_PM_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557037_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557038_s_at3 - -
          Human Genome U95 Set 42731_at - -
          Human Genome U95 Set 42731_PM_at - -
          Human Genome U95 Set 68187_at - -
          Human Genome U95 Set 68187_PM_at - -
          Human Genome U95 Set 72537_PM_at - -
          Human Genome U95 Set 72537_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A049778 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A049778 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 198011 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0