Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00419899_m1
          See other LINGO1-AS1 GE Assays ›
          Gene Symbol:
          LINGO1-AS1
          Gene Name:
          LINGO1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 77641764 - 77649240 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00419899_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINGO1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045123.1 2 - 3 322 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500007946.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000715.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772242 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772254 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268242 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30268209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09772248 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3464749 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 62186 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3464749 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1562748_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR15006404.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006404.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006404.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15006404.hg.1 TC15000715.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15006407.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006407.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15006407.hg.1 TC15000715.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15006406.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006406.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006406.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15006406.hg.1 TC15000715.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15006405.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006405.hg.1 NR_045123.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15006405.hg.1 TC15000715.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500156943.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156943.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156943.hg.1 TC1500007946.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500156943.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156943.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156944.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156944.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156944.hg.1 TC1500007946.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500156944.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156944.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156949.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156949.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156949.hg.1 TC1500007946.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500156949.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156949.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156948.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156948.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156948.hg.1 TC1500007946.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500156948.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156948.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156947.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156947.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156947.hg.1 TC1500007946.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500156947.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156947.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000558887.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007946.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156945.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156945.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156945.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156945.hg.1 TC1500007946.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500156945.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156945.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156945.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156942.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156942.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156942.hg.1 NR_045123.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500156942.hg.1 TC1500007946.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500156942.hg.1 ENST00000558691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156942.hg.1 ENST00000560265.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156942.hg.1 NR_045123.1 Refseq
          Human Genome U133 Plus 2.0 Array 1562748_at - -
          Human Genome U133 Plus 2.0 Array 1562748_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562748_at11 - -
          Human Genome U95 Set 62377_at - -
          Human Genome U95 Set 62377_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053111 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053111 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 373518 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0