Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00431103_m1
          See other NUP35 GE Assays ›
          Gene Symbol:
          NUP35
          Gene Name:
          nucleoporin 35
          Gene Aliases:
          MP-44, MP44, NP44, NUP53
          Chromosome Location:
          Chr.2: 183117490 - 183161684 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00431103_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NUP35
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001287584.1 NP_001274513.1 6 - 7 743 136
          NM_001287585.1 NP_001274514.1 6 - 7 763 136
          NM_138285.4 NP_612142.2 5 - 6 640 136
          XM_006712254.2 XP_006712317.1 5 - 6 605 136
          XM_011510576.2 XP_011508878.1 7 - 8 871 136
          XM_011510577.2 XP_011508879.1 7 - 8 1058 136
          XM_017003308.1 XP_016858797.1 5 - 6 796 136
          GenBank mRNA AF411516.1 - 5 - 6 640 136
          AF514993.1 - 5 - 6 597 136
          AK290760.1 - 5 - 6 552 136
          AK298199.1 - 6 - 7 673 136
          BC035597.1 - 5 - 6 564 136
          BC047029.1 - 5 - 6 627 5541023 136
          BC061698.1 - 5 - 6 594 4132440 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200016578.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001093.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NUP35

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          mRNA export from nucleus
          tRNA export from nucleus
          NLS-bearing protein import into nucleus
          nuclear pore organization
          mitotic nuclear membrane disassembly
          regulation of glucose transmembrane transport
          viral process
          protein sumoylation
          viral transcription
          gene silencing by RNA
          transmembrane transport
          intracellular transport of virus
          regulation of cellular response to heat
          nucleotide binding
          single-stranded DNA binding
          protein binding
          phospholipid binding
          structural constituent of nuclear pore

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02016370.hg.1 ENST00000295119.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02016370.hg.1 ENST00000409798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02016370.hg.1 ENST00000541912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02016370.hg.1 NM_138285.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02016370.hg.1 TC02001093.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 ENST00000295119.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 ENST00000374930.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 ENST00000409798.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 ENST00000479162.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 NM_001287584.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 NM_001287585.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 NM_138285.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 NR_109856.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 TC0200016578.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 ENST00000295119.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 ENST00000374930.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 ENST00000409798.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 ENST00000479162.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 NM_001287584.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 NM_001287585.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 NM_138285.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200224456.hg.1 NR_109856.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 171687 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0