Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00536319_s1
          See other ARHGAP5-AS1 GE Assays ›
          Gene Symbol:
          ARHGAP5-AS1
          Gene Name:
          ARHGAP5 antisense RNA 1 (head to head)
          Gene Aliases:
          C14orf128
          Chromosome Location:
          Chr.14: 32075419 - 32076699 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00536319_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARHGAP5-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027263.1 1 1166 99
          GenBank mRNA AI493138.1 - 1 99
          BC007251.1 - 1 1029 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW207880.1 - 1 Chr. 14 - 32075448 - 32075929 [+] on Build GRCh38 99
          GenBankmRNA AA972693.1 - 1 Chr. 14 - 32075424 - 32075863 [+] on Build GRCh38 99
          GenBankmRNA BX279651.1 - 1 Chr. 14 - 32075424 - 32075832 [+] on Build GRCh38 99
          GenBankmRNA CA312469.1 - 1 Chr. 14 - 32075424 - 32076228 [+] on Build GRCh38 99
          GenBankmRNA T71206.1 - 1 Chr. 14 - 32075415 - 32075660 [+] on Build GRCh38 99
          GenBankmRNA CF139324.1 - 1 Chr. 14 - 32075341 - 32075755 [+] on Build GRCh38 99
          GenBankmRNA BP395809.1 - 1 269 Chr. 14 - 32075419 - 32075801 [-] on Build GRCh38 99
          GenBankmRNA DB541571.1 - 1 348 Chr. 14 - 32075420 - 32075881 [-] on Build GRCh38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream nucleotide binding protein like NUBPL C14orf127,
          IND1,
          huInd1
          -214.2kb Anti-sense Chr. 14 - 31561385 - 31861293 on NCBI Build GRCh38
          Downstream Rho GTPase activating protein 5 ARHGAP5 GFI2,
          RhoGAP5,
          p190-B,
          p190BRhoGAP
          577.0 Anti-sense Chr. 14 - 32076152 - 32159728 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29663982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29663978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29663973 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29663983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29663976 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09206236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29663981 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 151635 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 6884 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 55283 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 19432 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 758738 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3348362 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P25746 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P25746 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 26755 - -
          Agilent Human Genome CGH Microarray 44B A_14_P202178 - -
          Agilent Human Genome CGH Microarray 44B A_14_P137450 - -
          Agilent Human Genome CGH Microarray 44A A_14_P137450 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 40641 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 44974 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P202178 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P25746 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3348362 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P25746 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3348362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664024 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663990 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663995 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664000 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663995 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664000 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663990 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29663994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664024 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29664042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09206264 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P25746 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P202178 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P137450 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P137450 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P202178 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P202178 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P137450 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 7664 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 28300 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 12146 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P137450 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P202178 - -
          Agilent Human 2 cDNA Microarray (G4101A) 4335140 - -
          Agilent Human 2 cDNA Microarray (G4101A) 4335140 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206260 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691607 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206242 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206252 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206250 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206228 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691592 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691603 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206246 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206237 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206266 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206256 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206240 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206248 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206230 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206280 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691598 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206272 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691602 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691594 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206283 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691601 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206277 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206284 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206275 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691591 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206263 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691593 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206244 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206286 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16691596 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206257 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206278 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09206235 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P202178 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P137450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7978540 - -
          Affymetrix Human Gene 1.0 ST Array 7978541 - -
          Affymetrix Human Gene 1.0 ST Array 7978539 - -
          Affymetrix Human Gene 1.1 ST Array 7978538 - -
          Affymetrix HT HG-U133+ PM Array Plate 228889_PM_at - -
          Human Genome U133 Plus 2.0 Array 228889_at - -
          Human Genome U133 Plus 2.0 Array 228889_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228889_at3 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050690 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050690 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 187637 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1804908 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1804908 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1804908 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0