Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019060.hg.1 |
ENST00000318672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019060.hg.1 |
ENST00000396865.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019060.hg.1 |
ENST00000416547.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019060.hg.1 |
ENST00000512595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019060.hg.1 |
ENST00000515629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019060.hg.1 |
ENST00000539768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019060.hg.1 |
NM_173514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019060.hg.1 |
TC05001365.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000318672.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000396865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000416547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000503817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000503891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000505708.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000508124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000511233.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000512208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000512595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000514806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000515629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NM_001258286.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NM_001258287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NM_001282429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NM_173514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NR_047649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
TC0500010779.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000396865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000318672.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000416547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000503817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000503891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000505708.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000508124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000511233.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000512208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000512595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000514806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
ENST00000515629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NM_001258286.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NM_001258287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NM_001282429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NM_173514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175193.hg.1 |
NR_047649.1 |
Refseq |