Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00544489_s1
          See other HIST1H2AJ GE Assays ›
          Gene Symbol:
          HIST1H2AJ
          Gene Name:
          histone cluster 1, H2aj
          Gene Aliases:
          H2A/E, H2AFE, dJ160A22.4
          Chromosome Location:
          Chr.6: 27814302 - 27814740 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00544489_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HIST1H2AJ
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021066.2 NP_066544.1 1 359 85
          GenBank mRNA BC133048.1 - 1 448 40147070 85
          BC133050.1 - 1 448 40147072 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600011225.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001422.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HIST1H2AJ

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24820190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24820193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04630042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24820194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24820192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04630050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24820191 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 130894 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 22171 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3344086 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P394510 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P168014 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 42564 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 13275 - -
          Agilent Human Genome CGH Microarray 44B A_14_P120961 - -
          Agilent Human Genome CGH Microarray 44A A_14_P120961 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 27126 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P120961 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3344086 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3344086 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820192 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P168014 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P394510 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P120961 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04630050 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04630048 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04630044 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04630051 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04630042 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04630047 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P120961 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P120961 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P394510 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P168014 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 5458 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 6896 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 3692 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 9962 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P120961 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630005 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630021 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630012 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630022 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630048 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630047 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04629999 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630042 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630018 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630009 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630024 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630051 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630015 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630036 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630006 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630000 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630011 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630034 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630014 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630003 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04629997 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630029 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P120961 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_021066_at - -
          Affymetrix Human Gene 1.0 ST Array 8124519 - -
          Affymetrix Human Gene 1.0 ST Array 8124520 - -
          Affymetrix Human Gene 1.1 ST Array 8124518 - -
          Affymetrix HT HG-U133+ PM Array Plate 208583_PM_x_at - -
          Affymetrix Human Genome U219 Array 11715117_PM_x_at - -
          Affymetrix Human Genome U219 Array 11715117_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06016339.hg.1 NM_021066.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06016339.hg.1 TC06001422.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600175708.hg.1 NM_021066.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600175708.hg.1 TC0600011225.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600175708.hg.1 NM_021066.2 Refseq
          Human Genome U133 Plus 2.0 Array 207156_PM_at - -
          Human Genome U133 Plus 2.0 Array 214542_x_at - -
          Human Genome U133 Plus 2.0 Array 208583_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 207156_at - -
          Human Genome U133 Plus 2.0 Array 214554_at - -
          Human Genome U133 Plus 2.0 Array 208583_x_at - -
          Human Genome U133 Plus 2.0 Array 214542_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 214554_PM_at - -
          Human Genome U133A 2.0 Array 214554_PM_at - -
          Human Genome U133A 2.0 Array 208583_PM_x_at - -
          Human Genome U133A 2.0 Array 207156_at - -
          Human Genome U133A 2.0 Array 207156_PM_at - -
          Human Genome U133A 2.0 Array 214542_x_at - -
          Human Genome U133A 2.0 Array 208583_x_at - -
          Human Genome U133A 2.0 Array 214554_at - -
          Human Genome U133A 2.0 Array 214542_PM_x_at - -
          AffymetrixPathwork Diagnostics Pathchip pw511996 - -
          Human Genome U95 Set 34157_PM_f_at - -
          Human Genome U95 Set 285_g_at - -
          Human Genome U95 Set 285_PM_g_at - -
          Human Genome U95 Set 34156_i_at - -
          Human Genome U95 Set 34157_f_at - -
          Human Genome U95 Set 284_at - -
          Human Genome U95 Set 34156_PM_i_at - -
          Human Genome U95 Set 31768_PM_at - -
          Human Genome U95 Set 31768_at - -
          Human Genome U95 Set 284_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208583_x_at7 - -
          Human Genome U133 Set 214554_at - -
          Human Genome U133 Set 208583_PM_x_at - -
          Human Genome U133 Set 207156_PM_at - -
          Human Genome U133 Set 214554_PM_at - -
          Human Genome U133 Set 214542_PM_x_at - -
          Human Genome U133 Set 208583_x_at - -
          Human Genome U133 Set 214542_x_at - -
          Human Genome U133 Set 207156_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029568 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029568 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 122554 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1755318 BC066237.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1755318 BC133048.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1755318 BC133050.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1755318 NM_021066.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1755318 BC066232.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1755318 BC066237.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1755318 BC133048.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1755318 BC133050.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1755318 BC066232.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1755318 BC066237.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1755318 BC133048.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1755318 BC133050.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1755318 NM_021066.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1755318 BC066232.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1755318 BC066237.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1755318 BC133048.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1755318 BC133050.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1755318 BC066232.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1755318 BC066237.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1755318 BC133048.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1755318 BC133050.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1755318 BC066232.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_15718715-S NM_021066.2 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_10419-S - -
          Illumina human-6 v1.0 expression beadchip 0004120390 - -
          Illumina humanRef-8 v1.0 expression beadchip 0004120390 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1755318 BC066237.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1755318 BC133048.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1755318 BC133050.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1755318 BC066232.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1755318 BC066237.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1755318 BC133048.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1755318 BC133050.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1755318 BC066232.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 2100465 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_10419 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1755318 BC066237.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1755318 BC133048.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1755318 BC133050.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1755318 BC066232.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1755318 BC066237.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1755318 BC133048.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1755318 BC133050.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1755318 BC066232.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1755318 BC066237.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1755318 BC133048.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1755318 BC133050.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1755318 BC066232.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1755318 BC066237.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1755318 BC133048.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1755318 BC133050.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1755318 BC066232.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1755318 BC066237.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1755318 BC133048.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1755318 BC133050.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1755318 NM_021066.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1755318 BC066232.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0