Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00559361_m1
          See other TSPO GE Assays ›
          Gene Symbol:
          TSPO
          Gene Name:
          translocator protein
          Gene Aliases:
          BPBS, BZRP, DBI, IBP, MBR, PBR, PBS, PKBS, PTBR, mDRC, pk18
          Chromosome Location:
          Chr.22: 43151514 - 43163242 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00559361_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TSPO
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000714.5 NP_000705.2 2 - 3 298 70
          NM_001256530.1 NP_001243459.1 2 - 3 493 70
          NM_001256531.1 NP_001243460.1 2 - 3 402 70
          GenBank mRNA AF075589.1 - 1 - 2 101 70
          AF075590.1 - 1 - 2 101 70
          AY998017.1 - 1 - 2 193 70
          BC001110.2 - 2 - 3 248 2989070 70
          BG749252.1 - 2 - 3 403 70
          BG750685.1 - 2 - 3 69
          BG752086.1 - 2 - 3 299 70
          BP420304.1 - 2 - 3 278 70
          BQ057964.1 - 283 70
          BT006949.1 - 1 - 2 178 70
          CR456409.1 - 1 - 2 187 70
          M36035.1 - 2 - 3 239 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200007547.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000362.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TSPO

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to mitochondrion
          steroid biosynthetic process
          heme biosynthetic process
          anion transport
          chloride transport
          lipid transport
          apoptotic process
          signal transduction
          chemical synaptic transmission
          aging
          steroid metabolic process
          cell population proliferation
          glial cell migration
          response to xenobiotic stimulus
          response to manganese ion
          response to vitamin B1
          positive regulation of necrotic cell death
          peripheral nervous system axon regeneration
          adrenal gland development
          negative regulation of protein ubiquitination
          regulation of cholesterol transport
          response to progesterone
          negative regulation of tumor necrosis factor production
          response to testosterone
          positive regulation of apoptotic process
          negative regulation of nitric oxide biosynthetic process
          behavioral response to pain
          regulation of steroid biosynthetic process
          positive regulation of mitochondrial depolarization
          positive regulation of calcium ion transport
          contact inhibition
          positive regulation of glial cell proliferation
          negative regulation of glial cell proliferation
          cellular response to lipopolysaccharide
          cellular response to zinc ion
          cellular hypotonic response
          maintenance of protein location in mitochondrion
          negative regulation of autophagy of mitochondrion
          negative regulation of ATP metabolic process
          positive regulation of reactive oxygen species metabolic process
          androgen binding
          protein binding
          benzodiazepine receptor activity
          cholesterol binding
          transmembrane transporter binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22006946.hg.1 TC22000362.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22006946.hg.1 ENST00000329563.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006946.hg.1 ENST00000337554.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006946.hg.1 ENST00000396265.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22006946.hg.1 NM_000714.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000472378.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000472378.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000472378.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000472378.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153518.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153518.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153518.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153518.hg.1 TC2200007547.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200153518.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153518.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153518.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000472378.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000472378.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000472378.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000472378.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_000714.5 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007547.hg.1 NR_046308.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 TC2200007547.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153517.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 NM_000714.5 Refseq
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 NR_046308.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 TC2200007547.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000329563.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000337554.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000396265.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000428336.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 ENST00000583777.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 NM_000714.5 Refseq
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 NM_001256530.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 NM_001256531.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200153522.hg.1 NR_046308.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0