Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017542.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017542.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017542.hg.1 |
TC04001283.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017543.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017543.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017543.hg.1 |
TC04001283.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017544.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017544.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017544.hg.1 |
TC04001283.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0400174876.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174876.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174876.hg.1 |
TC0400011014.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400174876.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174876.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174877.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174877.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174877.hg.1 |
TC0400011014.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400174877.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174877.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174875.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174875.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174875.hg.1 |
TC0400011014.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400174875.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174875.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174879.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174879.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174879.hg.1 |
TC0400011014.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400174879.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174879.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
ENST00000296027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011014.hg.1 |
NM_002994.4 |
Refseq |