Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00608036_m1
          See other CDK6 GE Assays ›
          Gene Symbol:
          CDK6
          Gene Name:
          cyclin dependent kinase 6
          Gene Aliases:
          MCPH12, PLSTIRE
          Chromosome Location:
          Chr.7: 92604921 - 92836627 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00608036_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDK6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145306.1 NP_001138778.1 3 - 4 876 139
          NM_001259.6 NP_001250.1 3 - 4 771 139
          XM_006715835.2 XP_006715898.1 3 - 4 996 139
          GenBank mRNA AK313491.1 - 3 - 4 814 139
          BC052264.1 - 3 - 4 752 6377987 139
          X66365.1 - 2 - 3 476 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700011785.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001603.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CDK6

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          positive regulation of cell-matrix adhesion
          type B pancreatic cell development
          protein phosphorylation
          Notch signaling pathway
          negative regulation of cell population proliferation
          response to virus
          regulation of gene expression
          positive regulation of gene expression
          astrocyte development
          dentate gyrus development
          lateral ventricle development
          T cell differentiation in thymus
          gliogenesis
          cell dedifferentiation
          negative regulation of cell differentiation
          negative regulation of myeloid cell differentiation
          regulation of erythrocyte differentiation
          negative regulation of monocyte differentiation
          negative regulation of osteoblast differentiation
          negative regulation of cell cycle
          positive regulation of fibroblast proliferation
          generation of neurons
          negative regulation of epithelial cell proliferation
          cell division
          hematopoietic stem cell differentiation
          cellular response to phorbol 13-acetate 12-myristate
          regulation of cell motility
          negative regulation of cellular senescence
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding
          cyclin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07024327.hg.1 ENST00000265734.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07024327.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07024327.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07024327.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07024327.hg.1 TC07001603.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000491250.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000491250.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay PSR0700184223.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184223.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184223.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184223.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay PSR0700184223.hg.1 TC0700011785.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700184223.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184223.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184223.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184223.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000491250.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000491250.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0