Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00609608_m1
          See other ALOX15 GE Assays ›
          Gene Symbol:
          ALOX15
          Gene Name:
          arachidonate 15-lipoxygenase
          Gene Aliases:
          12-LOX, 15-LOX-1, 15LOX-1
          Chromosome Location:
          Chr.17: 4630919 - 4641665 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00609608_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ALOX15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001140.3 NP_001131.3 8 - 9 1176 72
          GenBank mRNA AK290309.1 - 8 - 9 1189 72
          AK296792.1 - 6 - 7 833 72
          AK296821.1 - 9 - 10 1261 72
          AK296904.1 - 8 - 9 1072 72
          AK312557.1 - 8 - 9 1189 72
          AK316126.1 - 8 - 9 1332 72
          BC029032.1 - 8 - 9 1176 5179700 72
          M23892.1 - 8 - 9 1165 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700009520.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001027.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Human CYP450 and other Oxygenases 1700 Inventoried MicroArray Quality Control (MAQC) Human CYP450 + oxygenases Human_CYP450_and_other_Oxygenases Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ALOX15

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          negative regulation of adaptive immune response
          phosphatidylethanolamine biosynthetic process
          leukotriene metabolic process
          inflammatory response
          positive regulation of cell-substrate adhesion
          arachidonic acid metabolic process
          lipoxygenase pathway
          bone mineralization
          positive regulation of actin filament polymerization
          positive regulation of heterotypic cell-cell adhesion
          response to endoplasmic reticulum stress
          regulation of peroxisome proliferator activated receptor signaling pathway
          cellular response to interleukin-13
          wound healing
          apoptotic cell clearance
          hepoxilin biosynthetic process
          oxidation-reduction process
          positive regulation of ERK1 and ERK2 cascade
          cellular response to calcium ion
          regulation of engulfment of apoptotic cell
          lipoxin A4 biosynthetic process
          arachidonate 12-lipoxygenase activity
          iron ion binding
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          hepoxilin-epoxide hydrolase activity
          arachidonate 15-lipoxygenase activity
          hepoxilin A3 synthase activity
          eoxin A4 synthase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000293761.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000570836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000572265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000572265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000572265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000572265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000572265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000573740.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000573740.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000573740.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000573740.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000573740.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000573740.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000574640.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576394.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576394.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576394.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576394.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576572.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576572.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576572.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576572.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576572.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 ENST00000576572.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009520.hg.1 NM_001140.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 ENST00000621574.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006705.hg.1 NR_045985.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900007970.hg.1 ENST00000340858.4 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 139446 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7f58f9545f-46b6w:80/100.66.130.62:80.
          git-commit: c36c52929dfaa9f2956040e6abd275853d6e1b95
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline