Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00610318_m1
          See other TGFBR1 GE Assays ›
          Gene Symbol:
          TGFBR1
          Gene Name:
          transforming growth factor beta receptor 1
          Gene Aliases:
          AAT5, ACVRLK4, ALK-5, ALK5, ESS1, LDS1, LDS1A, LDS2A, MSSE, SKR4, TGFR-1, tbetaR-I
          Chromosome Location:
          Chr.9: 99104038 - 99154192 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00610318_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TGFBR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004612.3 NP_004603.1 2 - 3 453 92
          XM_011518949.2 XP_011517251.1 2 - 3 308 92
          GenBank mRNA AJ619020.1 - 1 - 2 157 92
          AK302059.1 - 2 - 3 453 92
          AK302234.1 - 2 - 3 326 92
          L11695.1 - 2 - 3 412 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900008202.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000500.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Homo sapiens Human IL-2 Gene Expression in Activated and Quiescent T-Cell HUMAN COLORECTAL CANCER METASTASIS HUMAN MOLECULAR MECHANISMS OF CANCER HUMAN SMAD SIGNALING NETWORK HUMAN PKA SIGNALING HUMAN P53 MEDIATED APOPTOSIS HUMAN FACTORS PROMOTING CARDIOGENESIS IN VERTEBRATES HUMAN PANCREATIC ADENOCARCINOMA Inventoried Human_IL-2_Gene_Expression_in_Activated_and_Quiescent_T-Cells Human_Pancreatic_Adenocarcinoma Human_Factors_Promoting_Cardiogenesis_in_Vertebrates Human_PKA_Signaling Human_Molecular_Mechanisms_of_Cancer Human_p53_Mediated_Apoptosis Human_Colorectal_Cancer_Metastasis Human_SMAD_Signaling_Network Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TGFBR1

          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          skeletal system development
          angiogenesis
          in utero embryonic development
          kidney development
          blastocyst development
          epithelial to mesenchymal transition
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lens development in camera-type eye
          ventricular trabecula myocardium morphogenesis
          ventricular compact myocardium morphogenesis
          proepicardium development
          regulation of transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          cell cycle arrest
          signal transduction
          transforming growth factor beta receptor signaling pathway
          heart development
          positive regulation of cell proliferation
          germ cell migration
          male gonad development
          post-embryonic development
          anterior/posterior pattern specification
          regulation of epithelial to mesenchymal transition
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          signal transduction by protein phosphorylation
          collagen fibril organization
          positive regulation of cell growth
          positive regulation of cell migration
          negative regulation of transforming growth factor beta receptor signaling pathway
          regulation of protein ubiquitination
          negative regulation of chondrocyte differentiation
          activin receptor signaling pathway
          intracellular signal transduction
          wound healing
          endothelial cell activation
          extracellular structure organization
          regulation of protein binding
          endothelial cell migration
          positive regulation of transcription, DNA-templated
          thymus development
          neuron fate commitment
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          mesenchymal cell differentiation
          artery morphogenesis
          cell motility
          positive regulation of cellular component movement
          positive regulation of filopodium assembly
          positive regulation of stress fiber assembly
          positive regulation of protein kinase B signaling
          parathyroid gland development
          palate development
          pharyngeal system development
          regulation of cardiac muscle cell proliferation
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          positive regulation of SMAD protein import into nucleus
          ventricular septum morphogenesis
          coronary artery morphogenesis
          response to cholesterol
          cellular response to transforming growth factor beta stimulus
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of occluding junction disassembly
          positive regulation of apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type I
          receptor signaling protein activity
          type II transforming growth factor beta receptor binding
          protein binding
          ATP binding
          growth factor binding
          SMAD binding
          metal ion binding
          transforming growth factor beta binding
          I-SMAD binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374990.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000374994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000546584.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000546584.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000546584.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000547314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000547314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000547314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000547314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000548365.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000548365.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000548365.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000548365.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549021.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549021.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549021.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549021.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549021.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000549766.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000550253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552573.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552573.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 ENST00000552573.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001130916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_001306210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008202.hg.1 NM_004612.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009436.hg.1 ENST00000594355.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009436.hg.1 ENST00000594355.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009436.hg.1 ENST00000594355.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009436.hg.1 ENST00000594355.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009436.hg.1 ENST00000594355.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009436.hg.1 ENST00000594355.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0