Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00610607_m1
          See other TACC2 GE Assays ›
          Gene Symbol:
          TACC2
          Gene Name:
          transforming acidic coiled-coil containing protein 2
          Gene Aliases:
          AZU-1, ECTACC
          Chromosome Location:
          Chr.10: 121989174 - 122254545 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00610607_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TACC2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006997.3 NP_008928.1 10 - 11 2857 96
          NM_206861.2 NP_996743.1 13 - 14 2922 96
          NM_206862.3 NP_996744.3 16 - 17 8484 96
          XM_005269388.1 XP_005269445.1 17 - 18 8401 96
          XM_005269389.4 XP_005269446.1 17 - 18 8925 96
          XM_005269390.1 XP_005269447.1 18 - 19 8344 96
          XM_005269391.1 XP_005269448.1 16 - 17 8341 96
          XM_005269393.1 XP_005269450.1 17 - 18 8308 96
          XM_005269394.1 XP_005269451.1 16 - 17 8275 96
          XM_005269395.1 XP_005269452.1 16 - 17 8266 96
          XM_005269397.1 XP_005269454.1 15 - 16 8140 96
          XM_006717548.1 XP_006717611.1 17 - 18 8296 96
          XM_011539126.1 XP_011537428.1 17 - 18 8398 96
          XM_011539128.1 XP_011537430.1 15 - 16 2974 96
          XM_011539129.1 XP_011537431.1 14 - 15 2839 96
          XM_011539130.1 XP_011537432.1 14 - 15 2746 96
          XM_011539131.1 XP_011537433.1 10 - 11 2812 96
          XM_011539132.1 XP_011537434.1 11 - 12 2755 96
          XM_017015484.1 XP_016870973.1 17 - 18 8389 96
          XM_017015488.1 XP_016870977.1 15 - 16 8035 96
          XM_017015491.1 XP_016870980.1 10 - 11 2952 96
          XM_017015492.1 XP_016870981.1 10 - 11 2859 96
          XM_017015496.1 XP_016870985.1 11 - 12 2753 96
          XM_017015497.1 XP_016870986.1 11 - 12 3012 96
          XM_017015498.1 XP_016870987.1 10 - 11 2877 96
          GenBank mRNA AF095791.2 - 10 - 11 2444 96
          AF528098.1 - 13 - 14 2903 96
          AF528099.1 - 16 - 17 8465 96
          AL833304.1 - 15 - 16 7583 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000009140.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000883.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TACC2

          Gene Ontology Categories:

          Function(s)

          microtubule cytoskeleton organization
          cell population proliferation
          cerebral cortex development

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000260733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000334433.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000340076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000358010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000360561.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000368999.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000369000.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000369001.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000369004.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000369005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000453444.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000513429.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000515273.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 ENST00000515603.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 NM_006997.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 NM_206860.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 NM_206861.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 NM_206862.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010729.hg.1 TC10000883.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000260733.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000334433.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000358010.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000368999.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000369005.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000513429.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 NM_006997.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 NM_206861.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 NM_206862.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 TC1000009140.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000260733.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000334433.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000358010.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000368999.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000369005.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 ENST00000513429.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 NM_006997.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 NM_206861.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162593.hg.1 NM_206862.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d58dc5f8-fsfvr:80/100.66.134.75:80.
          git-commit: 38b8a393e477ae4d9d89cc5f7a29948d21ae29e2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc