Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00738473_m1
          See other KIRREL3-AS3 GE Assays ›
          Gene Symbol:
          KIRREL3-AS3
          Gene Name:
          KIRREL3 antisense RNA 3
          Gene Aliases:
          NCRNA00288, PRR10
          Chromosome Location:
          Chr.11: 127002910 - 127006058 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00738473_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KIRREL3-AS3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040078.1 1 - 2 1412 138
          GenBank mRNA AK097543.1 - 1 - 2 1413 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009463.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001179.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532075 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104282 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104308 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16514527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104281 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104290 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16514526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104289 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532072 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104284 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08104295 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28532068 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129763 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 57036 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 27042 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 26432 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 14753 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 38937 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P201515 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P389770 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P111759 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P201515 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P389770 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 109293 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 43447 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 19861 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 87620 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P111759 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P201515 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P111759 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P201515 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7945127 - -
          Affymetrix Human Gene 1.0 ST Array 7945129 - -
          Affymetrix Human Gene 1.0 ST Array 7945126 - -
          Affymetrix Human Gene 1.0 ST Array 7945128 - -
          Affymetrix Human Gene 1.1 ST Array 7945125 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553478_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11014895.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11014895.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11014895.hg.1 TC11001179.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11014892.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11014892.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11014892.hg.1 TC11001179.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11014889.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11014889.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11014889.hg.1 TC11001179.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11014893.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11014893.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11014893.hg.1 TC11001179.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11014894.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11014894.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11014894.hg.1 TC11001179.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 ENST00000404882.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009463.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11014890.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11014890.hg.1 NR_040078.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11014890.hg.1 TC11001179.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100171215.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171215.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100171215.hg.1 TC1100009463.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100171215.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100171215.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171218.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171218.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100171218.hg.1 TC1100009463.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100171218.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171218.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100171217.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171217.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100171217.hg.1 TC1100009463.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100171217.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171217.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100171219.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171219.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100171219.hg.1 TC1100009463.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100171219.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171219.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100171216.hg.1 ENST00000404882.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171216.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171216.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100171216.hg.1 TC1100009463.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100171216.hg.1 ENST00000404882.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171216.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100171216.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 ENST00000404882.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 ENST00000525678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 NR_040078.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009463.hg.1 NR_040078.1 Refseq
          Human Genome U133 Plus 2.0 Array 1553478_at - -
          Human Genome U133 Plus 2.0 Array 1553478_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553478_at1 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047127 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047127 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 227303 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1696399 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1794703 NR_040078.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1794703 AK097543.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1696399 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1696399 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1794703 NR_040078.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1794703 AK097543.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1794703 NR_040078.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1794703 AK097543.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0