Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00742077_s1
          See other SLC25A3P1 GE Assays ›
          Gene Symbol:
          SLC25A3P1
          Gene Name:
          solute carrier family 25 member 3 pseudogene 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 53438370 - 53440020 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00742077_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC25A3P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK097753.1 - 1 1990 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15061335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231175 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231180 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231221 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20231223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00203859 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 60513 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 12606 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3309365 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3749 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P936189 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3748 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 36511 - -
          Agilent Human Genome CGH Microarray 44B A_14_P120254 - -
          Agilent Human Genome CGH Microarray 44A A_14_P120254 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20924 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3309365 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3309365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203866 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203857 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15061334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231206 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203861 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231208 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15061335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231204 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203861 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15061334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231208 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231204 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15061335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203857 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231206 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20231210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00203866 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P936189 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P120254 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P120254 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P120254 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P936189 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 29023 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 29779 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P120254 - -
          Agilent Human 1B Microarray G4111A A_32_P936189 - -
          Agilent Human 1B Microarray G4111A A_32_P458356 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203833 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203881 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203861 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203844 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203879 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203858 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203874 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15061328 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15061326 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203835 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203840 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203838 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203854 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203869 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15061335 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15061336 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203849 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203863 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203824 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203829 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203856 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15061334 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203865 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203851 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203884 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203871 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203847 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203826 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203842 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203831 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15061332 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203822 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203837 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203867 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00203882 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P120254 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1560944_PM_at - -
          Human Genome U133 Plus 2.0 Array 1560944_at - -
          Human Genome U133 Plus 2.0 Array 1560944_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560944_at1 - -
          Human Genome U95 Set 86987_PM_at - -
          Human Genome U95 Set 86987_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045900 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045900 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3249524 NR_002314.3 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3249524 AK097753.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3249524 BC128116.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3249524 BC128117.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3249524 NR_002314.3 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3249524 AK097753.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3249524 BC128116.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3249524 BC128117.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3249524 NR_002314.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3249524 AK097753.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3249524 BC128116.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3249524 BC128117.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3249524 NR_002314.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3249524 AK097753.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3249524 BC128116.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3249524 BC128117.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix