Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00742161_s1
          See other ERAS GE Assays ›
          Gene Symbol:
          ERAS
          Gene Name:
          ES cell expressed Ras
          Gene Aliases:
          HRAS2, HRASP
          Chromosome Location:
          Chr.X: 48826513 - 48829869 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00742161_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ERAS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AB093575.1 - 1 1218 138

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741931 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741932 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32449420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32449436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32449430 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32449404 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32449437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11741935 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 131007 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 57551 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P314585 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P314585 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 19577 - -
          Agilent Human Genome CGH Microarray 44B A_14_P110723 - -
          Agilent Human Genome CGH Microarray 44A A_14_P110723 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43320 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59999 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P41671065 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P314585 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P314585 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741947 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741943 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449420 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449404 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741943 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449420 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449404 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741947 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32449436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11741948 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P314585 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P110723 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741942 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741950 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741930 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741945 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741938 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741953 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741939 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741932 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741936 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741941 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11741948 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341544 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341550 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11741941 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341552 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11741938 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341548 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341553 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341545 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11741939 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11741940 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341547 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341549 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341543 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341546 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17341551 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P110723 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P110723 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P314585 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 1214 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 30188 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 23780 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P110723 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03702791 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741942 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17341546 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741974 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741983 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741953 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741979 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741976 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741965 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17341551 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741972 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741970 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741938 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741948 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741950 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17341549 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741941 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741939 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741936 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741968 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17341544 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17341542 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17341569 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741930 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741932 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741981 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741945 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17341543 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741967 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11741934 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P110723 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_181532_at - -
          Affymetrix Human Gene 1.0 ST Array 8167410 - -
          Affymetrix Human Gene 1.0 ST Array 8167408 - -
          Affymetrix Human Gene 1.0 ST Array 8167411 - -
          Affymetrix Human Gene 1.0 ST Array 8167409 - -
          Affymetrix Human Gene 1.1 ST Array 8167407 - -
          Affymetrix Human Genome U219 Array 11738569_at - -
          Affymetrix Human Genome U219 Array 11738569_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0X00007206.hg.1 ENST00000338270.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007206.hg.1 ENST00000338270.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007206.hg.1 ENST00000338270.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007206.hg.1 ENST00000338270.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007206.hg.1 NM_181532.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007206.hg.1 NM_181532.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007206.hg.1 NM_181532.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007206.hg.1 ENST00000338270.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007206.hg.1 ENST00000338270.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007206.hg.1 ENST00000338270.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007206.hg.1 ENST00000338270.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007206.hg.1 NM_181532.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007206.hg.1 NM_181532.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007206.hg.1 NM_181532.3 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040962 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040962 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 199780 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1721386 NM_181532.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1721386 AB093575.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1721386 BC101642.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1721386 BC101644.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1721386 AB093575.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1721386 BC101642.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1721386 BC101644.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1721386 NM_181532.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1721386 AB093575.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1721386 BC101642.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1721386 BC101644.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1721386 AB093575.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1721386 BC101642.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1721386 BC101644.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1721386 AB093575.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1721386 BC101642.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1721386 BC101644.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31581555-S NM_181532.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31581555-S AB093575.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_18435-S - -
          Illumina human-6 v1.0 expression beadchip 0006380673 - -
          Illumina humanRef-8 v1.0 expression beadchip 0006380673 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1721386 AB093575.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1721386 BC101642.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1721386 BC101644.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1721386 AB093575.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1721386 BC101642.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1721386 BC101644.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 6420347 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_18435 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1721386 AB093575.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1721386 BC101642.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1721386 BC101644.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1721386 AB093575.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1721386 BC101642.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1721386 BC101644.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1721386 AB093575.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1721386 BC101642.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1721386 BC101644.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1721386 AB093575.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1721386 BC101642.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1721386 BC101644.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1721386 NM_181532.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1721386 AB093575.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1721386 BC101642.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1721386 BC101644.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0