Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00748620_s1
          See other AKR7A2 GE Assays ›
          Gene Symbol:
          AKR7A2
          Gene Name:
          aldo-keto reductase family 7 member A2
          Gene Aliases:
          AFAR, AFAR1, AFB1-AR1, AKR7
          Chromosome Location:
          Chr.1: 19302708 - 19312146 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00748620_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AKR7A2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320979.1 NP_001307908.1 6 1180 114
          NM_003689.3 NP_003680.2 7 1285 114
          GenBank mRNA AF026947.1 - 7 1253 114
          AK225267.1 - 6 848 114
          BC004111.2 - 7 1212 3832140 114
          BC007352.2 - 7 1260 114
          BC011586.1 - 7 1212 4620484 114
          BC012171.1 - 7 1260 4642811 114
          BC013996.2 - 7 1216 3625337 114
          BU944847.1 - 1 356 114
          EU794591.1 - 7 1285 114
          Y16675.1 - 7 1247 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013131.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002298.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AKR7A2

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          cellular aldehyde metabolic process
          xenobiotic metabolic process
          daunorubicin metabolic process
          doxorubicin metabolic process
          alditol:NADP+ 1-oxidoreductase activity
          aldo-keto reductase (NADP) activity
          electron transfer activity
          phenanthrene-9,10-epoxide hydrolase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00010623.hg.1 ENST00000445747.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010623.hg.1 ENST00000445747.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010623.hg.1 ENST00000445747.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035227.hg.1 ENST00000235835.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035227.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035227.hg.1 TC01002298.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01035226.hg.1 ENST00000235835.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035226.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035226.hg.1 TC01002298.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01035225.hg.1 ENST00000235835.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035225.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035225.hg.1 TC01002298.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035228.hg.1 ENST00000235835.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035228.hg.1 NM_003689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035228.hg.1 TC01002298.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 TC0100013131.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197226.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010623.hg.1 ENST00000445747.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010623.hg.1 ENST00000445747.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010623.hg.1 ENST00000445747.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 ENST00000492217.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013131.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100197223.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197223.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197223.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197223.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100197223.hg.1 TC0100013131.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100197223.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197223.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197223.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197223.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100197224.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197224.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197224.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197224.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100197224.hg.1 TC0100013131.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100197224.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197224.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197224.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197224.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 TC0100013131.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 ENST00000481966.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 ENST00000489286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197225.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100197222.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197222.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197222.hg.1 NM_003689.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100197222.hg.1 TC0100013131.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100197222.hg.1 ENST00000235835.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197222.hg.1 ENST00000330072.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197222.hg.1 NM_003689.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 150154 - -
          ABI Human Genome Survey Microarray Version 2.0 226673 - -
          ABI Human Genome Survey Microarray Version 2.0 235331 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0