Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00753146_s1
          See other FLJ37201 GE Assays ›
          Gene Symbol:
          FLJ37201
          Gene Name:
          tigger transposable element derived 2 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 89691300 - 89697928 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00753146_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ37201
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026835.1 2 3497 81
          GenBank mRNA AK094520.1 - 1 2651 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000011385.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16371417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384432 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384433 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384401 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384396 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16371415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384428 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384411 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384399 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16371418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07384421 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27773681 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 132089 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 854 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 646271 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 606073 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 606838 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P246091 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P246091 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 36680 - -
          Agilent Human Genome CGH Microarray 44B A_14_P120203 - -
          Agilent Human Genome CGH Microarray 44A A_14_P120203 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14114 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P120203 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P246091 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P246091 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384465 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773704 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773706 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384461 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384462 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773710 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384466 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773706 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384466 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384465 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773710 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384461 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773704 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07384462 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27773718 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P246091 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P120203 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P120203 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P120203 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 169193 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 66668 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P120203 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10842825 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P32716149 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384417 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384446 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384450 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384404 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384455 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16371422 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384453 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384431 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384409 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16371419 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384459 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384424 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384476 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384469 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384448 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384430 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16371420 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384400 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384484 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384433 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384465 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384414 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384478 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384443 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384435 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384461 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384473 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384463 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16371425 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384480 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384427 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384402 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384406 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384487 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16371427 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384418 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384482 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384467 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16371416 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384438 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384411 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384441 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384422 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384457 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384489 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07384398 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P10842825 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P120203 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7934962 - -
          Affymetrix Human Gene 1.0 ST Array 7934963 - -
          Affymetrix Human Gene 1.0 ST Array 7934964 - -
          Affymetrix Human Gene 1.0 ST Array 7934965 - -
          Affymetrix Human Gene 1.1 ST Array 7934961 - -
          Affymetrix HT HG-U133+ PM Array Plate 1560654_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 ENST00000455699.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 ENST00000455699.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177373.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177373.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177373.hg.1 TC1000011385.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177373.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177373.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 ENST00000449882.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 ENST00000455699.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 ENST00000455699.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011385.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177372.hg.1 NR_026835.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000177372.hg.1 TC1000011385.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177372.hg.1 NR_026835.1 Refseq
          Human Genome U133 Plus 2.0 Array 1560654_PM_at - -
          Human Genome U133 Plus 2.0 Array 1560654_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560654_at11 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A043091 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A043091 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 680492 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1727099 NR_026835.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1727099 AK094520.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1727099 NR_026835.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1727099 AK094520.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1727099 NR_026835.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1727099 AK094520.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727099 NR_026835.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727099 AK094520.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0