Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00827245_g1
          See other LOC494141 GE Assays ›
          Gene Symbol:
          LOC494141
          Gene Name:
          solute carrier family 25 member 51 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 18209138 - 18213578 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00827245_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC494141
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC026082.1 - 1 - 2 54 73
          BC056262.1 - 1 - 2 58 73
          DB462574.1 - 1 - 2 58 73

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07671192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28071050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28071079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16424904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28071081 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07671187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28071049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16424903 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28071055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07671188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28071065 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07671190 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 99246 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 82727 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 723393 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230726 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 391752 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4665 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4666 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02402912 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P112512 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28071065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671188 - -
          Agilent Human 1B Microarray G4111A A_32_P177270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671187 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671193 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671188 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671189 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16424873 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671183 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671195 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671181 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16424875 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671192 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671190 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671191 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P11013245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7938754 - -
          Affymetrix Human Gene 1.0 ST Array 7938751 - -
          Affymetrix Human Gene 1.0 ST Array 7938753 - -
          Affymetrix Human Gene 1.0 ST Array 7938755 - -
          Affymetrix Human Gene 1.0 ST Array 7938752 - -
          Affymetrix Human Gene 1.1 ST Array 7938750 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046190 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046192 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046191 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046192 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046190 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046191 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 241085 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3292784 BC026082.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3292784 NR_026541.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0