Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00832119_s1
          See other LINC00851 GE Assays ›
          Gene Symbol:
          LINC00851
          Gene Name:
          long intergenic non-protein coding RNA 851
          Gene Aliases:
          DZANK1-AS1
          Chromosome Location:
          Chr.20: 18379049 - 18381484 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00832119_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00851
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034167.1 2 534 74
          GenBank mRNA BC033985.1 - 2 539 5295734 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY002680.1 - 2 535 Chr. 20 - 18379049 - 18381339 [+] on Build GRCh38 74
          GenBankmRNA AA815297.1 - 1 Chr. 20 - 18381207 - 18381484 [-] on Build GRCh38 74
          GenBankmRNA AA723764.1 - 1 Chr. 20 - 18381125 - 18381477 [-] on Build GRCh38 74
          GenBankmRNA DA765599.1 - 1 Chr. 20 - 18381009 - 18381589 [-] on Build GRCh38 74
          GenBankmRNA BI461247.1 - 2 539 Chr. 20 - 18379049 - 18381464 [+] on Build GRCh38 74
          GenBankmRNA AA917035.1 - 1 Chr. 20 - 18381206 - 18381484 [-] on Build GRCh38 74
          GenBankmRNA BI560072.1 - 2 539 Chr. 20 - 18379049 - 18381478 [+] on Build GRCh38 74
          GenBankmRNA AI138545.1 - 1 Chr. 20 - 18381016 - 18381484 [-] on Build GRCh38 74
          GenBankmRNA AW134562.1 - 1 Chr. 20 - 18381076 - 18381484 [-] on Build GRCh38 74
          GenBankmRNA AW273805.1 - 1 Chr. 20 - 18380979 - 18381484 [-] on Build GRCh38 74
          GenBankmRNA AK096438.1 - 1 Chr. 20 - 18379442 - 18381589 [-] on Build GRCh38 74
          GenBankmRNA AA897427.1 - 1 Chr. 20 - 18381147 - 18381482 [-] on Build GRCh38 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105372550 LOC105372550 -21.5kb Anti-sense Chr. 20 - 18313725 - 18359309 on NCBI Build GRCh38
          Downstream double zinc ribbon and ankyrin repeat domains 1 DZANK1 ANKRD64,
          C20orf12,
          C20orf84,
          bA189K21.1,
          bA189K21.8,
          dJ568F9.2
          1.9kb Anti-sense Chr. 20 - 18383367 - 18467185 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000006826.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000121.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31739808 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03494620 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8061168 - -
          Affymetrix Human Gene 1.1 ST Array 8061167 - -
          Affymetrix HT HG-U133+ PM Array Plate 231447_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2000006826.hg.1 ENST00000423033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006826.hg.1 ENST00000594642.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006826.hg.1 NR_034167.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001566.hg.1 TC20000121.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20001566.hg.1 ENST00000423033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001566.hg.1 NR_034167.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006826.hg.1 ENST00000423033.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006826.hg.1 ENST00000594642.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006826.hg.1 NR_034167.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000146706.hg.1 ENST00000423033.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146706.hg.1 ENST00000594642.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146706.hg.1 NR_034167.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000146706.hg.1 TC2000006826.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000146706.hg.1 NR_034167.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000146706.hg.1 ENST00000423033.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146706.hg.1 ENST00000594642.1 Ensembl
          Human Genome U133 Plus 2.0 Array 231447_at - -
          Human Genome U133 Plus 2.0 Array 231447_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231447_at5 - -
          Human Genome U95 Set 81466_at - -
          Human Genome U95 Set 81466_PM_at - -
          Human Genome U133 Set 231447_at - -
          Human Genome U133 Set 231447_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018090 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018090 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 224366 - -
          ABI Human Genome Survey Microarray Version 2.0 229399 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1678234 NR_034167.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1678234 BC033985.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1678234 NR_034167.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1678234 BC033985.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1678234 NR_034167.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1678234 BC033985.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1678234 NR_034167.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1678234 BC033985.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1678234 NR_034167.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1678234 BC033985.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1678234 NR_034167.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1678234 BC033985.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1678234 NR_034167.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1678234 BC033985.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1678234 NR_034167.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1678234 BC033985.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1678234 NR_034167.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1678234 BC033985.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc