Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000735.hg.1 |
ENST00000349637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000735.hg.1 |
ENST00000406844.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000735.hg.1 |
ENST00000447027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000735.hg.1 |
ENST00000529681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000735.hg.1 |
ENST00000546052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000735.hg.1 |
NM_002458.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000735.hg.1 |
TC11003510.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100145384.hg.1 |
ENST00000529681.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100145384.hg.1 |
NM_002458.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145384.hg.1 |
TC1100006509.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100145384.hg.1 |
NM_002458.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145384.hg.1 |
ENST00000529681.5 |
Ensembl |