Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00862300_m1
          See other LOC401445 GE Assays ›
          Gene Symbol:
          LOC401445
          Gene Name:
          uncharacterized LOC401445
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00862300_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC401445
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK057888.1 - 1 - 2 149 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101927815 LOC101927815 -144.7kb Sense Chr. 8 - 2530092 - 2728451 on NCBI Build GRCh38
          Downstream CUB and Sushi multiple domains 1 CSMD1 PPP1R24 57.3kb Sense Chr. 8 - 2935353 - 4995035 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192800 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192931 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16056505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192806 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919105 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919120 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192836 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919260 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919116 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192831 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192809 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16056502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919259 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192834 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192838 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919106 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192835 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192888 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05919261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26192946 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 411440 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 376396 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 906916 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 209161 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3469438 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 11246 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3469438 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38283544 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01850797 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01850797 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 689728 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline