Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00863353_g1
          See other RPL7L1 GE Assays ›
          Gene Symbol:
          RPL7L1
          Gene Name:
          ribosomal protein L7 like 1
          Gene Aliases:
          dJ475N16.4
          Chromosome Location:
          Chr.6: 42879163 - 42889896 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00863353_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RPL7L1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198486.3 NP_940888.3 1 - 2 336 139
          NR_134562.1 1 - 2 336 139
          XM_017010780.1 XP_016866269.1 1 - 2 789 139
          GenBank mRNA HY089482.1 - 1 - 2 337 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600014127.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000586.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_198486.3, XM_017010780.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RPL7L1

          Gene Ontology Categories:

          Function(s) Process(es)

          maturation of LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          blastocyst formation
          cytoplasmic translation
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06006657.hg.1 ENST00000304734.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006657.hg.1 ENST00000424341.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006657.hg.1 ENST00000483998.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006657.hg.1 ENST00000493763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006657.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06006657.hg.1 TC06000586.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800007667.hg.1 ENST00000520784.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011072.hg.1 ENST00000446889.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012812.hg.1 ENST00000437739.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010829.hg.1 ENST00000590816.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006652.hg.1 ENST00000304734.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006652.hg.1 ENST00000424341.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006652.hg.1 ENST00000493763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006652.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06006652.hg.1 TC06000586.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06006660.hg.1 ENST00000304734.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006660.hg.1 ENST00000424341.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006660.hg.1 ENST00000483998.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006660.hg.1 ENST00000493763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006660.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06006660.hg.1 TC06000586.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300012592.hg.1 ENST00000509986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011593.hg.1 ENST00000511702.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006897.hg.1 ENST00000425129.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008673.hg.1 ENST00000446092.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014796.hg.1 ENST00000454884.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009644.hg.1 ENST00000469911.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011090.hg.1 ENST00000470927.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010512.hg.1 ENST00000449851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010512.hg.1 ENST00000449851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000487619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000487619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000487619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000492836.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000492836.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000492836.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000497417.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000497417.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000497417.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000497417.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009189.hg.1 ENST00000572039.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011425.hg.1 ENST00000511857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010296.hg.1 ENST00000530523.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011798.hg.1 ENST00000513004.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009587.hg.1 ENST00000417594.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009587.hg.1 ENST00000423238.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015703.hg.1 ENST00000431298.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007751.hg.1 ENST00000466005.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007751.hg.1 ENST00000466005.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008075.hg.1 ENST00000455249.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009976.hg.1 ENST00000569728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009976.hg.1 ENST00000569728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014796.hg.1 ENST00000454884.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500007751.hg.1 ENST00000466005.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500007751.hg.1 ENST00000466005.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009587.hg.1 ENST00000423238.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007667.hg.1 ENST00000520784.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700008075.hg.1 ENST00000455249.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006897.hg.1 ENST00000425129.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 NM_198486.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 TC0600014127.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198754.hg.1 NM_198486.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011090.hg.1 ENST00000470927.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010512.hg.1 ENST00000449851.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010512.hg.1 ENST00000449851.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010296.hg.1 ENST00000530523.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009976.hg.1 ENST00000569728.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009976.hg.1 ENST00000569728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011798.hg.1 ENST00000513004.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011593.hg.1 ENST00000511702.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012812.hg.1 ENST00000437739.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008673.hg.1 ENST00000446092.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300009644.hg.1 ENST00000469911.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010829.hg.1 ENST00000590816.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011425.hg.1 ENST00000511857.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012592.hg.1 ENST00000509986.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011072.hg.1 ENST00000446889.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009189.hg.1 ENST00000572039.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015703.hg.1 ENST00000431298.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000397415.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000459829.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000462348.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000483998.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000487619.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000487619.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000487619.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000492836.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000492836.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000492836.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000497417.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000497417.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000497417.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000497417.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 ENST00000602561.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014127.hg.1 NM_198486.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009587.hg.1 ENST00000417594.2 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0