Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00864981_s1
          See other KCNJ3 GE Assays ›
          Gene Symbol:
          KCNJ3
          Gene Name:
          potassium voltage-gated channel subfamily J member 3
          Gene Aliases:
          GIRK1, KGA, KIR3.1
          Chromosome Location:
          Chr.2: 154698581 - 154858352 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00864981_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KCNJ3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001260508.1 NP_001247437.1 2 3251 69
          NM_002239.3 NP_002230.1 3 3468 69
          GenBank mRNA AK026384.1 - 1 218 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CK023806.1 - 1 204 Chr. 2 - 154856879 - 154857463 [+] on Build GRCh38 69
          GenBankmRNA BQ303941.1 - 1 106 Chr. 2 - 154856977 - 154857525 [+] on Build GRCh38 69
          GenBankmRNA BX488523.1 - 1 483 Chr. 2 - 154856598 - 154857235 [+] on Build GRCh38 69
          GenBankmRNA H91765.1 - 1 217 Chr. 2 - 154856864 - 154857310 [+] on Build GRCh38 69
          GenBankmRNA BQ303942.1 - 1 95 Chr. 2 - 154856988 - 154857530 [+] on Build GRCh38 69
          GenBankmRNA BG563492.1 - 1 195 Chr. 2 - 154856886 - 154857719 [+] on Build GRCh38 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC100144595 LOC100144595 -397.3kb Anti-sense Chr. 2 - 154435853 - 154457438 on NCBI Build GRCh38
          Downstream uncharacterized LOC101929378 LOC101929378 1.2mb Anti-sense Chr. 2 - 156020535 - 156254931 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200009707.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000947.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KCNJ3

          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transport
          regulation of ion transmembrane transport
          response to electrical stimulus
          potassium ion import across plasma membrane
          inward rectifier potassium channel activity
          protein binding
          G-protein activated inward rectifier potassium channel activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02013869.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02013869.hg.1 NM_002239.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02013869.hg.1 TC02000947.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000493505.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000493505.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000493505.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260510.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260510.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260510.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_001260510.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000493505.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000493505.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000493505.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260509.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260510.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260510.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260510.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_001260510.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009707.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200167796.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200167796.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200167796.hg.1 NM_001260508.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200167796.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200167796.hg.1 TC0200009707.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200167796.hg.1 NM_002239.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200167796.hg.1 ENST00000295101.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200167796.hg.1 ENST00000544049.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200167796.hg.1 NM_001260508.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 120159 - -
          ABI Human Genome Survey Microarray Version 2.0 300784 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0