Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015008.hg.1 |
NM_198455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015008.hg.1 |
TC07000992.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0700168310.hg.1 |
ENST00000378016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168310.hg.1 |
ENST00000378016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168310.hg.1 |
ENST00000623373.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168310.hg.1 |
NM_198455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168310.hg.1 |
TC0700009646.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700168310.hg.1 |
ENST00000378016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168310.hg.1 |
ENST00000378016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168310.hg.1 |
ENST00000623373.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168310.hg.1 |
NM_198455.2 |
Refseq |