Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00884256_s1
          See other ANP32AP1 GE Assays ›
          Gene Symbol:
          ANP32AP1
          Gene Name:
          acidic nuclear phosphoprotein 32 family member A pseudogene 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 35237326 - 35238063 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00884256_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANP32AP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003144.1 1 170 131
          GenBank mRNA AK311417.1 - 2 372 131
          DQ083510.1 - 1 170 131
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA693550.1 - 2 372 Chr. 15 - 35181799 - 35237703 [+] on Build GRCh38 131

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger protein 770 ZNF770 PRO1914 -248.9kb Anti-sense Chr. 15 - 34978341 - 34988296 on NCBI Build GRCh38
          Downstream diphthamine biosynthesis 6 DPH6 ATPBD4 66.7kb Anti-sense Chr. 15 - 35217356 - 35546222 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500006851.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000248.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070370 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09580046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070359 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070369 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09580057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070367 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09580073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070361 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09580051 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070364 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09580049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09580058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09580047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09580059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070363 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30070346 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P133670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580058 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16778336 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070369 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070364 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070375 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16778336 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580058 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070375 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070364 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070369 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30070365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09580073 - -
          Agilent Human 1B Microarray G4111A A_32_P133671 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580049 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580095 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580110 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580086 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580046 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580082 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580093 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580076 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580057 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580048 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580079 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580101 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580088 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580106 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580069 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580066 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16778328 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580090 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580072 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580070 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580104 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580099 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580108 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580059 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580051 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580053 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580061 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09580063 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7982532 - -
          Affymetrix Human Gene 1.1 ST Array 7982531 - -
          Affymetrix Human Transcriptome Array 2.0 PSR15001542.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15001542.hg.1 NR_003144.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15001542.hg.1 TC15000248.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500006851.hg.1 ENST00000559548.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006851.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006851.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006851.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006851.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006851.hg.1 NR_003144.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500147550.hg.1 ENST00000559548.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147550.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147550.hg.1 NR_003144.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500147550.hg.1 TC1500006851.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500147550.hg.1 NR_003144.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500147550.hg.1 ENST00000559548.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147550.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006851.hg.1 ENST00000559548.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500006851.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006851.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006851.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006851.hg.1 ENST00000560832.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006851.hg.1 NR_003144.1 Refseq
          Human Genome U133 Plus 2.0 Array 208571_PM_at - -
          Human Genome U133 Plus 2.0 Array 208571_at - -
          Human Genome U133A 2.0 Array 208571_PM_at - -
          Human Genome U133A 2.0 Array 208571_at - -
          Human Genome U133 Set 208571_PM_at - -
          Human Genome U133 Set 208571_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052744 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052744 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2056002 NR_003144.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2056002 DQ083510.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2056002 NR_003144.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2056002 DQ083510.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2056002 NR_003144.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2056002 DQ083510.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2056002 NR_003144.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2056002 DQ083510.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2056002 NR_003144.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2056002 DQ083510.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2056002 NR_003144.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2056002 DQ083510.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2056002 NR_003144.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2056002 DQ083510.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2056002 NR_003144.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2056002 DQ083510.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0