Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00893239_s1
          See other DBIL5P2 GE Assays ›
          Gene Symbol:
          DBIL5P2
          Gene Name:
          diazepam binding inhibitor-like 5 pseudogene 2
          Gene Aliases:
          ELP1P
          Chromosome Location:
          Chr.2: 63117851 - 63119542 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00893239_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DBIL5P2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036635.2 1 503 85
          GenBank mRNA AF229803.1 - 1 503 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream orthodenticle homeobox 1 OTX1 -60.0kb Anti-sense Chr. 2 - 63050057 - 63057831 on NCBI Build GRCh38
          Downstream WD repeat containing planar cell polarity effector WDPCP BBS15,
          C2orf86,
          CHDTHP,
          FRITZ,
          FRTZ
          1.2kb Sense Chr. 2 - 63120755 - 63840825 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200012791.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001903.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15249304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310759 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01229984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01229968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310754 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310765 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01229978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21310752 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 544637 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3893191 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P921906 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17233 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 35139 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3893191 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3893191 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P921906 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P35704886 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P921906 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35704886 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P35704886 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8052595 - -
          Affymetrix Human Gene 1.0 ST Array 8052596 - -
          Affymetrix Human Gene 1.0 ST Array 8052597 - -
          Affymetrix Human Gene 1.1 ST Array 8052594 - -
          Affymetrix HT HG-U133+ PM Array Plate 224258_PM_at - -
          Affymetrix Human Genome U219 Array 11762670_at - -
          Affymetrix Human Genome U219 Array 11762670_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02030454.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02030454.hg.1 NR_036635.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02030454.hg.1 TC02001903.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200012791.hg.1 ENST00000443827.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012791.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012791.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012791.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012791.hg.1 NR_036635.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012791.hg.1 NR_036635.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012791.hg.1 NR_036635.2 Refseq
          Affymetrix Human Clariom D Assay TC0200012791.hg.1 ENST00000443827.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012791.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012791.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012791.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012791.hg.1 NR_036635.2 Refseq
          Affymetrix Human Clariom D Assay TC0200012791.hg.1 NR_036635.2 Refseq
          Affymetrix Human Clariom D Assay TC0200012791.hg.1 NR_036635.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200193272.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193272.hg.1 NR_036635.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200193272.hg.1 TC0200012791.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193272.hg.1 NR_036635.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200193272.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193270.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193270.hg.1 NR_036635.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200193270.hg.1 TC0200012791.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193270.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193270.hg.1 NR_036635.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200193271.hg.1 ENST00000443827.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193271.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193271.hg.1 NR_036635.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200193271.hg.1 TC0200012791.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200193271.hg.1 ENST00000443827.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193271.hg.1 ENST00000491724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200193271.hg.1 NR_036635.2 Refseq
          Human Genome U133 Plus 2.0 Array 224258_at - -
          Human Genome U133 Plus 2.0 Array 224258_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224258_at1 - -
          Human Genome U95 Set 88896_PM_at - -
          Human Genome U95 Set 88896_at - -
          Human Genome U133 Set 224258_PM_at - -
          Human Genome U133 Set 224258_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013716 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013716 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 165958 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0