Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005997.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005997.hg.1 |
ENST00000361872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005997.hg.1 |
ENST00000373816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005997.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005997.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005997.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005997.hg.1 |
TC01000389.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000516968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000516968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007569.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
NM_024482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
TC0100007569.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
ENST00000373816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
ENST00000294409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
ENST00000361872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
ENST00000480454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
NM_006582.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153086.hg.1 |
NM_024482.2 |
Refseq |