Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00894885_g1
          See other AIF1 GE Assays ›
          Gene Symbol:
          AIF1
          Gene Name:
          allograft inflammatory factor 1
          Gene Aliases:
          AIF-1, IBA1, IRT-1, IRT1
          Chromosome Location:
          Chr.6: 31615209 - 31617025 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00894885_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AIF1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318970.1 NP_001305899.1 4 - 5 255 98
          NM_001623.4 NP_001614.3 4 - 5 316 98
          GenBank mRNA AF088038.1 - 4 - 5 262 98
          AK290771.1 - 4 - 5 251 98
          AV716806.1 - 4 - 5 236 98
          AV735626.1 - 4 - 5 362 98
          BC009474.1 - 4 - 5 295 4249306 98
          CD521003.1 - 4 - 5 223 98
          D86438.1 - 4 - 5 204 98
          EF070983.1 - 4 - 5 195 98
          JQ782414.1 - 4 - 5 290 98
          JQ782415.1 - 4 - 5 229 98
          U19713.1 - 4 - 5 219 98

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the JQ782415.1, NM_001318970.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AIF1

          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          positive regulation of protein phosphorylation
          phagocytosis, engulfment
          inflammatory response
          negative regulation of gene expression
          positive regulation of muscle hyperplasia
          Rac protein signal transduction
          actin filament polymerization
          cellular response to extracellular stimulus
          cellular response to hormone stimulus
          positive regulation of T cell proliferation
          negative regulation of apoptotic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of smooth muscle cell proliferation
          negative regulation of smooth muscle cell proliferation
          response to axon injury
          actin filament bundle assembly
          response to glucocorticoid
          response to electrical stimulus
          cellular response to morphine
          cellular response to interferon-gamma
          cellular response to hydroperoxide
          negative regulation of smooth muscle cell chemotaxis
          positive regulation of smooth muscle cell chemotaxis
          positive regulation of monocyte chemotaxis
          ruffle assembly
          positive regulation of G1/S transition of mitotic cell cycle
          positive regulation of T cell migration
          molecular_function
          calcium ion binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0